Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC310926 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025002.1 | NP_001020173.1 | 7 | 1174 | ||
GenBank mRNA | AY310143.1 | - | 7 | 1174 | ||
AY318956.1 | - | 7 | 1174 | |||
AY318962.1 | - | 7 | 1174 | |||
AY325183.1 | - | 7 | 1174 | |||
AY686708.1 | - | 1 | 52 |
Gene Symbol: Nek6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC160820.1 | - | 104 | 7459041 |