Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Agap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108230.1 | NP_001101700.1 | 10 | 1024 | ||
XM_006245396.2 | XP_006245458.2 | 12 | 5132 | |||
XM_006245397.2 | XP_006245459.2 | 12 | 5132 | |||
XM_006245398.2 | XP_006245460.2 | 12 | 5131 | |||
XM_006245400.2 | XP_006245462.2 | 11 | 5119 | |||
XM_006245401.2 | XP_006245463.2 | 12 | 5132 | |||
XM_006245402.2 | XP_006245464.2 | 12 | 5132 | |||
XM_006245403.2 | XP_006245465.2 | 11 | 5119 |