Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gdap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013201.1 | NP_001013219.1 | 12 | 1474 | ||
XM_006233032.2 | XP_006233094.1 | 13 | 1572 | |||
XM_006233033.2 | XP_006233095.1 | 12 | 1522 | |||
GenBank mRNA | BC082000.1 | - | 12 | 1474 | 7131173 | |
FQ211326.1 | - | 14 | 1655 |