Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aste1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040156.2 | NP_001035246.2 | 4 | 1854 | ||
XM_006243672.2 | XP_006243734.1 | 3 | 1609 | |||
XM_006243675.2 | XP_006243737.1 | 4 | 1501 | |||
XM_008766543.1 | XP_008764765.1 | 3 | 2038 | |||
XM_008766544.1 | XP_008764766.1 | 3 | 2038 | |||
XM_008766545.1 | XP_008764767.1 | 3 | 733 | |||
XM_008766546.1 | XP_008764768.1 | 3 | 379 | |||
XM_008766547.1 | XP_008764769.1 | 2 | 229 | |||
XM_008766548.1 | XP_008764770.1 | 3 | 273 | |||
GenBank mRNA | BC083913.1 | - | 4 | 401 | ||
BQ193960.1 | - | 2 | 567 |
Gene Symbol: Atp2c1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | M93018.1 | - | 2540 |