Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrfip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014269.1 | NP_001014291.1 | 8 | 2344 | ||
XM_006245444.2 | XP_006245506.1 | 22 | 3134 | |||
XM_006245445.2 | XP_006245507.1 | 22 | 3291 | |||
XM_006245446.2 | XP_006245508.1 | 21 | 3089 | |||
XM_006245447.2 | XP_006245509.1 | 21 | 3062 | |||
XM_006245448.2 | XP_006245510.1 | 21 | 3041 | |||
XM_006245450.2 | XP_006245512.1 | 20 | 2947 | |||
XM_006245451.2 | XP_006245513.1 | 19 | 2944 | |||
XM_006245452.2 | XP_006245514.1 | 19 | 2875 | |||
XM_006245455.2 | XP_006245517.1 | 11 | 2537 | |||
XM_006245456.2 | XP_006245518.1 | 11 | 2695 | |||
XM_006245457.2 | XP_006245519.1 | 10 | 2623 | |||
XM_006245458.2 | XP_006245520.1 | 8 | 2279 | |||
XM_008767285.1 | XP_008765507.1 | 21 | 3095 | |||
XM_008767286.1 | XP_008765508.1 | 12 | 2615 | |||
GenBank mRNA | BC081883.1 | - | 8 | 2344 | 7108398 |