Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: E2f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100778.1 | NP_001094248.1 | 6, 7 | 1210 | ||
XM_008762363.1 | XP_008760585.1 | 6, 7 | 1207 |