Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100910973 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FM045317.1 | - | 2 | 39 | ||
FM097063.1 | - | 2 | 57 | |||
FM098643.1 | - | 2 | 229 | |||
FM110991.1 | - | 2 | 32 | |||
FQ212263.1 | - | 2 | 53 | |||
FQ220597.1 | - | 2 | 177 | |||
FQ230677.1 | - | 2 | 53 | |||
FQ230710.1 | - | 2 | 53 | |||
FQ234947.1 | - | 2 | 49 |