Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025769.1 | NP_001020940.1 | 2 | 125 | ||
GenBank mRNA | BC097436.1 | - | 2 | 125 | 7455517 |