Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM2 SIRNA Assays ›
Gene Symbol:
Tpm2
Gene Name:
tropomyosin 2, beta
Gene Aliases:
-
Chromosome Location:
Chr.5: 59016616 - 59025971 on Build Rnor_6.0
UniGene ID:
Rn.17580
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS365652
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001024345.2
NP_001019516.1
3
492
NM_001301235.1
NP_001288164.1
3
492
XM_008763680.1
XP_008761902.1
3
832
XM_008763681.1
XP_008761903.1
3
832
XM_008763682.1
XP_008761904.1
2
210
GenBank mRNA
BC090009.1
-
3
413
7324151
CF110059.1
-
3
378
FQ214609.1
-
3
427
FQ214635.1
-
3
429
FQ214711.1
-
3
429
FQ214816.1
-
3
408
FQ214823.1
-
3
429
FQ214899.1
-
3
429
FQ215016.1
-
3
392
FQ215066.1
-
3
429
FQ215076.1
-
3
389
FQ215143.1
-
3
429
FQ215150.1
-
3
429
FQ215210.1
-
3
429
FQ215302.1
-
3
429
FQ215335.1
-
3
389
FQ215502.1
-
3
429
FQ215601.1
-
3
429
FQ215676.1
-
3
429
FQ215883.1
-
3
427
FQ215920.1
-
3
427
FQ216112.1
-
3
428
FQ216187.1
-
3
429
FQ216228.1
-
3
429
FQ216294.1
-
3
428
FQ216477.1
-
3
429
FQ216571.1
-
3
429
FQ216617.1
-
3
431
FQ216760.1
-
3
429
FQ216828.1
-
3
429
FQ216972.1
-
3
430
FQ217324.1
-
3
429
FQ217401.1
-
3
429
FQ217430.1
-
3
429
FQ217483.1
-
3
432
FQ217946.1
-
3
430
FQ219788.1
-
3
389
FQ219821.1
-
3
429
FQ220371.1
-
3
429
FQ220438.1
-
3
389
FQ220606.1
-
3
429
FQ220675.1
-
3
429
FQ223760.1
-
3
492
FQ224873.1
-
3
431
FQ224893.1
-
3
431
FQ229470.1
-
3
429
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009416, NM_001277876, NM_001277875, M81086, AK132162, and others.
Gene Ontology Categories:
Function(s)
Process(es)
regulation of ATPase activity
actin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
git-url: https://github.com/thermofisher/magellan-search
git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix