Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tc2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025152.1 | NP_001020323.1 | 5 | 422 | ||
XM_008764831.1 | XP_008763053.1 | 4 | 2085 | |||
XM_008764832.1 | XP_008763054.1 | 5 | 1734 | |||
XM_008764833.1 | XP_008763055.1 | 5 | 1981 | |||
XM_008764834.1 | XP_008763056.1 | 4 | 955 | |||
XM_008764835.1 | XP_008763057.1 | 5 | 986 | |||
XM_008764836.1 | XP_008763058.1 | 5 | 604 | |||
XM_008764837.1 | XP_008763059.1 | 5 | 949 | |||
XM_008764838.1 | XP_008763060.1 | 4 | 918 | |||
XM_008764839.1 | XP_008763061.1 | 4 | 484 | |||
XM_008764840.1 | XP_008763062.1 | 3 | 351 | |||
GenBank mRNA | BC088432.1 | - | 5 | 422 | 7124658 | |
FQ211170.1 | - | 5 | 540 |