Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035255.1 | NP_001030332.1 | 3, 4 | 527 | ||
XM_006233540.2 | XP_006233602.1 | 4, 5 | 567 | |||
XM_006233541.2 | XP_006233603.1 | 4, 5 | 566 | |||
XM_006233542.2 | XP_006233604.1 | 4, 5 | 567 | |||
XM_006233543.2 | XP_006233605.1 | 5, 6 | 624 | |||
XM_008761567.1 | XP_008759789.1 | 4, 5 | 568 | |||
XM_008761568.1 | XP_008759790.1 | 5, 6 | 3615 | |||
XM_008761569.1 | XP_008759791.1 | 5, 6 | 631 | |||
XM_008761570.1 | XP_008759792.1 | 1, 2 | 815 | |||
GenBank mRNA | BC099157.1 | - | 3, 4 | 527 | 7381485 | |
FQ209707.1 | - | 4, 5 | 533 | |||
FQ234514.1 | - | 3, 4 | 542 |