Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srsf11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001035255.1 | NP_001030332.1 | 6 | 779 | ||
XM_006233540.2 | XP_006233602.1 | 7 | 819 | |||
XM_006233541.2 | XP_006233603.1 | 7 | 818 | |||
XM_006233542.2 | XP_006233604.1 | 7 | 819 | |||
XM_006233543.2 | XP_006233605.1 | 8 | 876 | |||
XM_008761567.1 | XP_008759789.1 | 7 | 820 | |||
XM_008761568.1 | XP_008759790.1 | 8 | 3867 | |||
XM_008761569.1 | XP_008759791.1 | 8 | 883 | |||
XM_008761570.1 | XP_008759792.1 | 4 | 1067 | |||
GenBank mRNA | BC099157.1 | - | 6 | 779 | 7381485 | |
FQ209707.1 | - | 7 | 785 | |||
FQ234514.1 | - | 6 | 794 |