Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017343.2 | NP_059039.2 | 2 | 217 | ||
GenBank mRNA | BC060577.1 | - | 2 | 216 | 6889406 | |
CF977112.1 | - | 2 | 221 | |||
D14688.1 | - | 2 | 187 | |||
EV769828.1 | - | 2 | 204 | |||
FQ215510.1 | - | 2 | 203 | |||
FQ229195.1 | - | 2 | 203 | |||
X52840.1 | - | 1 | 134 |
Gene Symbol: Myl12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135017.1 | NP_001128489.1 | 2 | 244 | ||
XM_006245667.2 | XP_006245729.1 | 2 | 398 | |||
XM_006245668.2 | XP_006245730.1 | 2 | 339 | |||
GenBank mRNA | EV767772.1 | - | 274 | |||
FQ215121.1 | - | 2 | 199 | |||
FQ215776.1 | - | 388 | ||||
FQ217558.1 | - | 2 | 174 | |||
FQ220614.1 | - | 230 | ||||
FQ223132.1 | - | 2 | 208 | |||
FQ228530.1 | - | 2 | 201 | |||
FQ229140.1 | - | 2 | 453 | |||
FQ232936.1 | - | 2 | 180 | |||
X05566.1 | - | 2 | 186 | |||
Z32519.1 | - | 2 | 186 |