Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Myl12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135017.1 | NP_001128489.1 | 4 | 617 | ||
XM_006245667.2 | XP_006245729.1 | 4 | 771 | |||
XM_006245668.2 | XP_006245730.1 | 4 | 712 | |||
GenBank mRNA | BQ207032.1 | - | 1 | 372 | ||
EV767772.1 | - | 647 | ||||
FQ212603.1 | - | 2 | 321 | |||
FQ215121.1 | - | 4 | 572 | |||
FQ215776.1 | - | 762 | ||||
FQ217399.1 | - | 4 | 663 | |||
FQ217558.1 | - | 4 | 547 | |||
FQ220614.1 | - | 603 | ||||
FQ223132.1 | - | 4 | 581 | |||
FQ228530.1 | - | 4 | 573 | |||
FQ228551.1 | - | 1 | 42 | |||
FQ229140.1 | - | 4 | 826 | |||
FQ232936.1 | - | 4 | 553 | |||
X05566.1 | - | 4 | 559 | |||
Z32519.1 | - | 4 | 559 |
Gene Symbol: Myl12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017343.2 | NP_059039.2 | 4 | 590 | ||
GenBank mRNA | BC060577.1 | - | 4 | 589 | 6889406 | |
D14688.1 | - | 4 | 560 | |||
EV769828.1 | - | 4 | 577 | |||
FQ215510.1 | - | 4 | 576 | |||
FQ229195.1 | - | 4 | 576 | |||
X52840.1 | - | 3 | 507 |