Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033961.1 | NP_001029133.1 | 5, 6 | 672 | ||
NM_020094.2 | NP_064479.2 | 7, 8 | 822 | |||
NM_020095.2 | NP_064480.2 | 6, 7 | 768 | |||
XM_008760448.1 | XP_008758670.1 | 5, 6 | 2380 | |||
XM_008760449.1 | XP_008758671.1 | 7, 8 | 931 | |||
XM_008760450.1 | XP_008758672.1 | 5, 6 | 572 | |||
GenBank mRNA | AF269283.1 | - | 7, 8 | 600 | ||
AF269284.1 | - | 6, 7 | 546 | |||
AF269285.1 | - | 5, 6 | 450 | |||
BC085905.1 | - | 5, 6 | 672 | 7193750 | ||
GU937871.1 | - | 5, 6 | 482 |
Gene Symbol: LOC100911951 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231581.2 | XP_006231643.1 | 5, 6 | 568 | ||
XM_008760479.1 | XP_008758701.1 | 7, 8 | 879 | |||
XM_008760480.1 | XP_008758702.1 | 6, 7 | 824 | |||
XM_008760481.1 | XP_008758703.1 | 5, 6 | 886 | |||
XM_008760482.1 | XP_008758704.1 | 5, 6 | 727 | |||
XM_008760483.1 | XP_008758705.1 | 7, 8 | 931 |