Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_171992.4 | NP_741989.3 | 3, 4 | 703 | ||
XM_008760168.1 | XP_008758390.1 | 3, 4 | 878 | |||
GenBank mRNA | AM258963.2 | - | 3, 4 | 756 | ||
CA510842.1 | - | 3, 4 | 704 | |||
D14014.1 | - | 3, 4 | 703 | |||
EV775485.1 | - | 2, 3 | 656 | |||
FQ231654.1 | - | 3, 4 | 716 | |||
X75207.1 | - | 3, 4 | 730 |