Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ERBB4 SIRNA Assays ›
          Gene Symbol:
          Erbb4
          Gene Name:
          v-erb-b2 avian erythroblastic leukemia viral oncogene 4
          Gene Aliases:
          -
          Chromosome Location:
          Chr.9: 75021790 - 76178936 on Build Rnor_6.0
          UniGene ID:
          Rn.163078
          Species:
          Rat
          Species Specific ID (Flybase ID):
          -
          siRNA ID RSS367698
          Size 20 nmol
          Availability Made To Order
          Catalog # 1330001
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Erbb4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021687.1 NP_067719.1 15 1836
          GenBank mRNA AF041838.1 - 15 1836
          AY375306.1 - 15 1803
          AY375307.1 - 15 1803
          AY375308.1 - 15 1803

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          neural crest cell migration
          transcription, DNA-templated
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          nervous system development
          heart development
          lactation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          embryonic pattern specification
          negative regulation of muscle cell apoptotic process
          cell migration
          peptidyl-tyrosine phosphorylation
          telencephalon development
          central nervous system morphogenesis
          olfactory bulb interneuron differentiation
          signal transduction by protein phosphorylation
          regulation of cell migration
          positive regulation of cell migration
          mammary gland development
          positive regulation of synaptic transmission, GABAergic
          response to progesterone
          odontogenesis
          response to drug
          positive regulation of tyrosine phosphorylation of Stat5 protein
          response to hydrogen peroxide
          negative regulation of apoptotic process
          surfactant homeostasis
          positive regulation of phosphatidylinositol 3-kinase activity
          mitochondrial fragmentation involved in apoptotic process
          cell fate commitment
          positive regulation of transcription, DNA-templated
          positive regulation of glucose import
          protein autophosphorylation
          positive regulation of epithelial cell proliferation
          positive regulation of cardiac muscle cell proliferation
          synapse maturation
          mammary gland epithelial cell differentiation
          mammary gland alveolus development
          cardiac muscle tissue regeneration
          positive regulation of ERK1 and ERK2 cascade
          positive regulation of STAT protein import into nucleus
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          receptor signaling protein tyrosine kinase activity
          epidermal growth factor receptor binding
          protein binding
          ATP binding
          protein homodimerization activity
          transcription regulatory region DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix