Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahsa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115034.1 | NP_001108506.1 | 2 | 318 | ||
GenBank mRNA | BC158698.1 | - | 2 | 275 | 9027714 | |
FQ223544.1 | - | 2 | 302 | |||
FQ232113.1 | - | 2 | 280 | |||
FQ233084.1 | - | 2 | 280 |