Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031110.1 | NP_112372.1 | 3, 4 | 216 | ||
GenBank mRNA | BC058465.1 | - | 3, 4 | 284 | 6920362 | |
FQ210967.1 | - | 3, 4 | 251 | |||
FQ217176.1 | - | 3, 4 | 239 | |||
FQ221188.1 | - | 3, 4 | 292 | |||
FQ221414.1 | - | 3, 4 | 243 | |||
FQ221793.1 | - | 3, 4 | 239 | |||
FQ221872.1 | - | 3, 4 | 239 | |||
FQ221910.1 | - | 3, 4 | 246 | |||
FQ222283.1 | - | 3, 4 | 268 | |||
FQ222285.1 | - | 3, 4 | 246 | |||
FQ222811.1 | - | 3, 4 | 240 | |||
FQ222934.1 | - | 3, 4 | 271 | |||
FQ222947.1 | - | 3, 4 | 239 | |||
FQ223175.1 | - | 3, 4 | 239 | |||
FQ223203.1 | - | 3, 4 | 241 | |||
FQ223895.1 | - | 3, 4 | 239 | |||
FQ229022.1 | - | 3, 4 | 239 | |||
K03250.1 | - | 3, 4 | 216 |