Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NCOA3 SIRNA Assays ›
Gene Symbol:
Ncoa3
Gene Name:
nuclear receptor coactivator 3
Gene Aliases:
Aib1, Tram-1
Chromosome Location:
Chr.3: 162692176 - 162788582 on Build Rnor_6.0
UniGene ID:
Rn.20691
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
RSS373161
Size
20 nmol
20 nmol
Availability
Made To Order
Catalog #
1330001
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ncoa3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_006235634.1
XP_006235696.1
4
459
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
male gonad development
positive regulation of gene expression
histone acetylation
positive regulation of cell growth
cellular response to hormone stimulus
positive regulation of intracellular steroid hormone receptor signaling pathway
positive regulation of intracellular estrogen receptor signaling pathway
multicellular organism growth
negative regulation of apoptotic process
positive regulation of keratinocyte differentiation
positive regulation of transcription from RNA polymerase II promoter
developmental growth
positive regulation of sequence-specific DNA binding transcription factor activity
vagina development
labyrinthine layer morphogenesis
mammary gland branching involved in thelarche
regulation of RNA biosynthetic process
RNA polymerase II regulatory region DNA binding
chromatin binding
transcription coactivator activity
histone acetyltransferase activity
signal transducer activity
ligand-dependent nuclear receptor binding
estrogen receptor binding
ligand-dependent nuclear receptor transcription coactivator activity
protein complex binding
nuclear hormone receptor binding
peroxisome proliferator activated receptor binding
protein dimerization activity
protein N-terminus binding
androgen receptor binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.32.0-2025.07.28-1.0