Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Elf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053520.2 | NP_445972.2 | 7 | 1450 | ||
XM_006252347.2 | XP_006252409.1 | 8 | 2733 | |||
XM_006252348.2 | XP_006252410.1 | 7 | 1471 | |||
XM_006252349.2 | XP_006252411.1 | 9 | 2899 | |||
XM_006252351.2 | XP_006252413.1 | 8 | 1977 | |||
XM_006252352.2 | XP_006252414.1 | 8 | 1937 | |||
XM_006252354.2 | XP_006252416.1 | 7 | 2625 | |||
XM_008770898.1 | XP_008769120.1 | 8 | 4237 | |||
XM_008770899.1 | XP_008769121.1 | 8 | 1933 | |||
XM_008770900.1 | XP_008769122.1 | 8 | 1489 | |||
XM_008770901.1 | XP_008769123.1 | 8 | 4213 | |||
XM_008770902.1 | XP_008769124.1 | 7 | 4129 | |||
XM_008770903.1 | XP_008769125.1 | 7 | 4105 | |||
GenBank mRNA | AB030216.1 | - | 6 | 1111 | ||
AB030217.1 | - | 5 | 946 | |||
AB046531.1 | - | 7 | 1229 | |||
FQ221456.1 | - | 9 | 2018 |