Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG6 SIRNA Assays ›
          Gene Symbol:
          Snhg6
          Gene Name:
          small nucleolar RNA host gene 6
          Gene Aliases:
          1110008H02Rik
          Chromosome Location:
          Chr.1: 9942025 - 9944118 on Build GRCm38
          UniGene ID:
          Mm.28311
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n251644

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Snhg6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024067.2 2 164
          GenBank mRNA AK003573.1 - 2 165
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA W71553.1 - 141 Chr. - - [-] on Build
          GenBankmRNA BY788907.1 - 2 163 Chr. 1 - 9942074 - 9944117 [-] on Build GRCm38
          GenBankmRNA CJ082230.1 - 2 163 Chr. 1 - 9942037 - 9944117 [-] on Build GRCm38
          GenBankmRNA BY043991.1 - 2 164 Chr. 1 - 9942072 - 9944117 [-] on Build GRCm38
          GenBankmRNA AA015539.1 - 2 124 Chr. 1 - 9942042 - 9944079 [-] on Build GRCm38
          GenBankmRNA AA023387.1 - 2 126 Chr. 1 - 9942023 - 9944083 [-] on Build GRCm38
          GenBankmRNA BY604390.1 - 2 169 Chr. 1 - 9942019 - 9944118 [-] on Build GRCm38
          GenBankmRNA BE853719.1 - 2 149 Chr. 1 - 9942153 - 9944103 [-] on Build GRCm38
          GenBankmRNA BI109518.1 - 2 147 Chr. 1 - 9942023 - 9944099 [-] on Build GRCm38
          GenBankmRNA AV468093.1 - 2 124 Chr. 1 - 9942028 - 9944077 [-] on Build GRCm38
          GenBankmRNA AA097989.1 - 2 134 Chr. 1 - 9942025 - 9944095 [-] on Build GRCm38
          GenBankmRNA BE134757.1 - 2 144 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
          GenBankmRNA W35693.1 - 93 Chr. - - [-] on Build
          GenBankmRNA W18398.1 - 133 Chr. - - [-] on Build
          GenBankmRNA BX523010.1 - 2 154 Chr. 1 - 9942023 - 9944107 [-] on Build GRCm38
          GenBankmRNA AA940467.1 - 2 143 Chr. 1 - 9941959 - 9944090 [-] on Build GRCm38
          GenBankmRNA BX516798.1 - 2 154 Chr. 1 - 9942023 - 9944105 [-] on Build GRCm38
          GenBankmRNA AL034794.1 - 1 254 Chr. 1 - 9942230 - 9942897 [-] on Build GRCm38
          GenBankmRNA AA002823.1 - 2 147 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
          GenBankmRNA AA289428.1 - 1 49 Chr. 1 - 9942025 - 9942695 [-] on Build GRCm38
          GenBankmRNA AA200900.1 - 2 128 Chr. 1 - 9942040 - 9944086 [-] on Build GRCm38
          GenBankmRNA AA155393.1 - 2 89 Chr. 1 - 9942025 - 9944043 [-] on Build GRCm38
          GenBankmRNA AA124208.1 - 2 148 Chr. 1 - 9942058 - 9944104 [-] on Build GRCm38
          GenBankmRNA BY346229.1 - 2 167 Chr. 1 - 9942118 - 9944121 [-] on Build GRCm38
          GenBankmRNA BY711012.1 - 2 163 Chr. 1 - 9941960 - 9944117 [-] on Build GRCm38
          GenBankmRNA CK793285.1 - 147 Chr. - - [-] on Build
          GenBankmRNA AV490092.1 - 2 128 Chr. 1 - 9942038 - 9944081 [-] on Build GRCm38
          GenBankmRNA BE624330.1 - 2 126 Chr. 1 - 9942023 - 9944080 [-] on Build GRCm38
          GenBankmRNA CJ320246.1 - 1 31 Chr. 1 - 9942026 - 9942683 [-] on Build GRCm38
          GenBankmRNA AV115890.2 - 343 Chr. - - [-] on Build
          GenBankmRNA AA250387.1 - 2 141 Chr. 1 - 9942054 - 9944087 [-] on Build GRCm38
          GenBankmRNA BY345355.1 - 2 167 Chr. 1 - 9942107 - 9944121 [-] on Build GRCm38
          GenBankmRNA BY111725.1 - 2 163 Chr. 1 - 9942091 - 9944117 [-] on Build GRCm38
          GenBankmRNA AV485517.1 - 2 151 Chr. 1 - 9942023 - 9944104 [-] on Build GRCm38
          GenBankmRNA AA023086.1 - 2 153 Chr. 1 - 9942024 - 9944105 [-] on Build GRCm38
          GenBankmRNA AA119805.1 - 2 139 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
          GenBankmRNA W40695.1 - 92 Chr. - - [-] on Build
          GenBankmRNA W54219.1 - 89 Chr. - - [-] on Build
          GenBankmRNA BY345466.1 - 2 167 Chr. 1 - 9942116 - 9944121 [-] on Build GRCm38
          GenBankmRNA W11261.1 - 147 Chr. - - [-] on Build
          GenBankmRNA BY034531.1 - 2 164 Chr. 1 - 9942060 - 9944117 [-] on Build GRCm38
          GenBankmRNA AA146116.1 - 2 84 Chr. 1 - 9942033 - 9944038 [-] on Build GRCm38
          GenBankmRNA W98878.1 - 142 Chr. - - [-] on Build
          GenBankmRNA AW988981.1 - 2 144 Chr. 1 - 9942023 - 9944098 [-] on Build GRCm38
          GenBankmRNA AA098159.1 - 2 141 Chr. 1 - 9942037 - 9944097 [-] on Build GRCm38
          GenBankmRNA AA014503.1 - 2 105 Chr. 1 - 9942168 - 9944058 [-] on Build GRCm38
          GenBankmRNA BY703829.1 - 2 163 Chr. 1 - 9942025 - 9944117 [-] on Build GRCm38
          GenBankmRNA BF715116.1 - 2 151 Chr. 1 - 9942023 - 9944105 [-] on Build GRCm38
          GenBankmRNA W54183.1 - 146 Chr. - - [-] on Build
          GenBankmRNA BY344080.1 - 2 168 Chr. 1 - 9942094 - 9944121 [-] on Build GRCm38
          GenBankmRNA AV115922.2 - 2 185 Chr. 1 - 9941973 - 9944118 [-] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix