Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zxda | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003292.1 | 1 | 4701 | |||
GenBank mRNA | AK078672.1 | - | 1 | 70 | ||
AK155431.1 | - | 2 | 3944 | |||
AK164166.1 | - | 1 | 1741 |