Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2900060B14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027901.1 | 1 | 93 | |||
GenBank mRNA | AK019334.1 | - | 1 | 94 | ||
AK019335.1 | - | 1 | 94 | |||
AK019348.1 | - | 1 | 94 | |||
AK050598.1 | - | 1 | 93 | |||
AK083371.1 | - | 1 | 93 | |||
AK169832.1 | - | 1 | 94 |