Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mir133b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029902.1 | 1 | 37 |
Gene Symbol: Linc-md1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | RNADB ID | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
GenBank mRNA | AK132542.1 | - | LIT3444 4831414J02Rik |
1 | 3786 | |
JN699052.1 | - | 3 | 368 |