Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SMIM4 SIRNA Assays ›
          Gene Symbol:
          Smim4
          Gene Name:
          small integral membrane protein 4
          Gene Aliases:
          2010107H07Rik, Gm41122
          Chromosome Location:
          Chr.14: 31124505 - 31128930 on Build GRCm38
          UniGene ID:
          Mm.379116
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n254717

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Smim4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA AK007443.1 - 1 269
          AK019298.1 - 1 244
          AK036279.1 - 232
          CX238554.1 - 251
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CB320048.1 - 250 Chr. - - [-] on Build
          GenBankmRNA DT908430.1 - 1 182 Chr. 14 - 31124506 - 31128844 [-] on Build GRCm38
          GenBankmRNA AV144712.1 - 1 154 Chr. 14 - 31124506 - 31128816 [-] on Build GRCm38
          GenBankmRNA BY259485.1 - 1 236 Chr. 14 - 31124506 - 31128898 [-] on Build GRCm38
          GenBankmRNA BU842352.1 - 1 232 Chr. 14 - 31124506 - 31128891 [-] on Build GRCm38
          GenBankmRNA AV451934.1 - 1 219 Chr. 14 - 31124505 - 31128884 [-] on Build GRCm38
          GenBankmRNA BY712428.1 - 1 244 Chr. 14 - 31128639 - 31128905 [-] on Build GRCm38
          GenBankmRNA AV053578.1 - 1 154 Chr. 14 - 31124506 - 31128816 [-] on Build GRCm38
          GenBankmRNA CA481214.1 - 270 Chr. - - [-] on Build
          GenBankmRNA BY708265.1 - 1 242 Chr. 14 - 31124653 - 31128903 [-] on Build GRCm38
          GenBankmRNA BY049007.1 - 1 247 Chr. 14 - 31124516 - 31128909 [-] on Build GRCm38
          GenBankmRNA CD350363.1 - 1 216 Chr. 14 - 31128303 - 31128878 [-] on Build GRCm38
          GenBankmRNA BY434381.1 - 1 241 Chr. 14 - 31124506 - 31128900 [-] on Build GRCm38
          GenBankmRNA BY076653.1 - 1 248 Chr. 14 - 31124505 - 31128908 [-] on Build GRCm38
          GenBankmRNA BY619590.1 - 1 239 Chr. 14 - 31124506 - 31128898 [-] on Build GRCm38
          GenBankmRNA BY070889.1 - 1 258 Chr. 14 - 31124525 - 31128919 [-] on Build GRCm38
          GenBankmRNA W54436.1 - 237 Chr. - - [-] on Build
          GenBankmRNA AA874321.1 - 1 238 Chr. 14 - 31124660 - 31128903 [-] on Build GRCm38
          GenBankmRNA BY615358.1 - 1 247 Chr. 14 - 31124506 - 31128898 [-] on Build GRCm38
          GenBankmRNA BU843299.1 - 250 Chr. - - [-] on Build
          GenBankmRNA BY254061.1 - 1 236 Chr. 14 - 31124506 - 31128898 [-] on Build GRCm38
          GenBankmRNA BB629120.1 - 1 229 Chr. 14 - 31088627 - 31128891 [-] on Build GRCm38

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Human: AK095910, BC017996, BG031013, AK000903.

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6bbccd986b-mx52t:80/100.66.130.62:80.
          git-commit: 717389b07c2b9f75290ff02e811d41bb86d4254a
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0