Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 2610001J05RIK SIRNA Assays ›
          Gene Symbol:
          2610001J05Rik
          Gene Name:
          RIKEN cDNA 2610001J05 gene
          Gene Aliases:
          C79299
          Chromosome Location:
          Chr.6: 13869074 - 13871483 on Build GRCm38
          UniGene ID:
          Mm.29615
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n254826

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 2610001J05Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_024619.1 3 465
          GenBank mRNA AK002763.1 - 3 448
          AK011259.1 - 3 448
          AK028169.1 - 3 466
          AK035027.1 - 3 448
          AK136158.1 - 3 449
          AK152937.1 - 501
          AW611020.1 - 3 466
          BC070405.1 - 2 432
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BY745481.1 - 3 501 Chr. 6 - 13870259 - 13871518 [-] on Build GRCm38
          GenBankmRNA BG086442.2 - 2 432 Chr. 6 - 13870281 - 13871147 [-] on Build GRCm38
          GenBankmRNA AA051696.1 - 2 424 Chr. 6 - 13870312 - 13871147 [-] on Build GRCm38
          GenBankmRNA BY703269.1 - 3 448 Chr. 6 - 13869949 - 13871465 [-] on Build GRCm38
          GenBankmRNA DV647729.1 - 2 432 Chr. 6 - 13870104 - 13871147 [-] on Build GRCm38
          GenBankmRNA DV056552.1 - 466 Chr. - - [-] on Build
          GenBankmRNA BI695701.1 - 1 296 Chr. 6 - 13869921 - 13870707 [-] on Build GRCm38
          GenBankmRNA BQ564552.1 - 2 351 Chr. 6 - 13870135 - 13871076 [-] on Build GRCm38
          GenBankmRNA BM939634.1 - 2 432 Chr. 6 - 13870208 - 13871147 [-] on Build GRCm38
          GenBankmRNA CA469415.1 - 3 484 Chr. 6 - 13870132 - 13871465 [-] on Build GRCm38
          GenBankmRNA BI685945.1 - 2 426 Chr. 6 - 13870231 - 13871147 [-] on Build GRCm38
          GenBankmRNA BQ178406.1 - 2 433 Chr. 6 - 13870132 - 13871147 [-] on Build GRCm38
          GenBankmRNA AW912235.1 - 2 432 Chr. 6 - 13870316 - 13871146 [-] on Build GRCm38
          GenBankmRNA AV103829.2 - 2 428 Chr. 6 - 13869800 - 13871154 [-] on Build GRCm38
          GenBankmRNA BX525698.1 - 2 407 Chr. 6 - 13870318 - 13871133 [-] on Build GRCm38
          GenBankmRNA BU694337.1 - 2 432 Chr. 6 - 13870238 - 13871147 [-] on Build GRCm38
          GenBankmRNA CK023323.1 - 3 443 Chr. 6 - 13870090 - 13871460 [-] on Build GRCm38
          GenBankmRNA BF151248.1 - 2 418 Chr. 6 - 13870343 - 13871144 [-] on Build GRCm38
          GenBankmRNA C79299.1 - 493 Chr. - - [-] on Build
          GenBankmRNA BE198626.1 - 2 432 Chr. 6 - 13870326 - 13871147 [-] on Build GRCm38
          GenBankmRNA CB590232.1 - 2 414 Chr. 6 - 13870029 - 13871140 [-] on Build GRCm38
          GenBankmRNA BB627264.1 - 3 448 Chr. 6 - 13870214 - 13871465 [-] on Build GRCm38
          GenBankmRNA CB849720.2 - 2 436 Chr. 6 - 13870345 - 13871141 [-] on Build GRCm38
          GenBankmRNA AA138847.1 - 2 359 Chr. 6 - 13870333 - 13871085 [-] on Build GRCm38
          GenBankmRNA BE198399.1 - 2 432 Chr. 6 - 13870319 - 13871147 [-] on Build GRCm38
          GenBankmRNA BU524555.1 - 3 450 Chr. 6 - 13870141 - 13871465 [-] on Build GRCm38
          GenBankmRNA CK020651.1 - 321 Chr. - - [-] on Build
          GenBankmRNA CX238014.1 - 3 446 Chr. 6 - 13870280 - 13871464 [-] on Build GRCm38
          GenBankmRNA AI785717.1 - 2 433 Chr. 6 - 13870255 - 13871147 [-] on Build GRCm38
          GenBankmRNA CF615966.1 - 1 338 Chr. 6 - 13869918 - 13870746 [-] on Build GRCm38
          GenBankmRNA CO803384.1 - 2 417 Chr. 6 - 13870134 - 13871137 [-] on Build GRCm38
          GenBankmRNA BY720110.1 - 3 449 Chr. 6 - 13870183 - 13871465 [-] on Build GRCm38
          GenBankmRNA CB596362.1 - 3 450 Chr. 6 - 13870150 - 13871465 [-] on Build GRCm38
          GenBankmRNA AA039161.1 - 3 448 Chr. 6 - 13870361 - 13871466 [-] on Build GRCm38
          GenBankmRNA CB600423.1 - 3 450 Chr. 6 - 13870148 - 13871465 [-] on Build GRCm38
          GenBankmRNA CV561544.1 - 2 418 Chr. 6 - 13870090 - 13871142 [-] on Build GRCm38
          GenBankmRNA BU935739.1 - 2 359 Chr. 6 - 13870044 - 13871465 [-] on Build GRCm38
          GenBankmRNA CF731670.1 - 2 364 Chr. 6 - 13870101 - 13871091 [-] on Build GRCm38
          GenBankmRNA BU512890.1 - 1 357 Chr. 6 - 13869912 - 13870742 [-] on Build GRCm38
          GenBankmRNA CB954743.1 - 300 Chr. - - [-] on Build
          GenBankmRNA AI182977.1 - 2 430 Chr. 6 - 13870402 - 13871147 [-] on Build GRCm38
          GenBankmRNA AW988917.1 - 2 401 Chr. 6 - 13870349 - 13871127 [-] on Build GRCm38
          GenBankmRNA BG173485.1 - 2 441 Chr. 6 - 13870103 - 13871147 [-] on Build GRCm38
          GenBankmRNA BU558791.1 - 3 450 Chr. 6 - 13870118 - 13871465 [-] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-75d45cbfdf-4zq6t:80/100.66.130.62:80.
          git-commit: a3c66eacce7c9fa3045440ddf4a86215532ee526
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix