Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 1110038B12RIK SIRNA Assays ›
          Gene Symbol:
          1110038B12Rik
          Gene Name:
          RIKEN cDNA 1110038B12 gene
          Gene Aliases:
          0610009C20Rik, 0910001G04Rik
          Chromosome Location:
          Chr.17: 34950235 - 34952471 on Build GRCm38
          UniGene ID:
          Mm.28895
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n255417

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 1110038B12Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015536.1 2, 3 156
          NR_027943.1 2, 3 153
          GenBank mRNA AK002372.1 - 2, 3 156
          AK003073.1 - 2, 3 154
          AK004150.1 - 2, 3 154
          BC058123.1 - 2, 3 140
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CB205178.1 - 2, 3 137 Chr. 17 - 34950231 - 34952452 [-] on Build GRCm38
          GenBankmRNA AW987897.1 - 2, 3 132 Chr. 17 - 34950243 - 34952447 [-] on Build GRCm38
          GenBankmRNA AV488693.1 - 2, 3 137 Chr. 17 - 34950268 - 34952455 [-] on Build GRCm38
          GenBankmRNA CB589057.1 - 2, 3 141 Chr. 17 - 34950231 - 34952456 [-] on Build GRCm38
          GenBankmRNA AV506956.1 - 2, 3 117 Chr. 17 - 34950337 - 34952428 [-] on Build GRCm38
          GenBankmRNA CB321306.1 - 140 Chr. - - [-] on Build
          GenBankmRNA CB588222.1 - 158 Chr. - - [-] on Build
          GenBankmRNA CF739395.1 - 2, 3 134 Chr. 17 - 34950241 - 34952449 [-] on Build GRCm38
          GenBankmRNA BX527841.1 - 2, 3 106 Chr. 17 - 34950248 - 34952420 [-] on Build GRCm38
          GenBankmRNA CB196482.1 - 2, 3 137 Chr. 17 - 34950231 - 34952454 [-] on Build GRCm38
          GenBankmRNA CB575170.1 - 138 Chr. - - [-] on Build
          GenBankmRNA BY307160.1 - 2, 3 152 Chr. 17 - 34950294 - 34952467 [-] on Build GRCm38
          GenBankmRNA BY054987.1 - 2, 3 154 Chr. 17 - 34950450 - 34952466 [-] on Build GRCm38
          GenBankmRNA AV505341.1 - 2, 3 130 Chr. 17 - 34950314 - 34952438 [-] on Build GRCm38
          GenBankmRNA CN694693.1 - 2, 3 142 Chr. 17 - 34951446 - 34952457 [-] on Build GRCm38
          GenBankmRNA BE859815.1 - 2, 3 124 Chr. 17 - 34950252 - 34952439 [-] on Build GRCm38
          GenBankmRNA BY702909.1 - 2, 3 156 Chr. 17 - 34950242 - 34952471 [-] on Build GRCm38
          GenBankmRNA AV479153.1 - 2, 3 120 Chr. 17 - 34950250 - 34952423 [-] on Build GRCm38
          GenBankmRNA AV005223.1 - 2, 3 166 Chr. 17 - 34951748 - 34952468 [-] on Build GRCm38
          GenBankmRNA BB867243.1 - 2, 3 167 Chr. 17 - 34950330 - 34952472 [-] on Build GRCm38
          GenBankmRNA W66707.1 - 106 Chr. - - [-] on Build
          GenBankmRNA BY086580.1 - 2, 3 150 Chr. 17 - 34950288 - 34952465 [-] on Build GRCm38
          GenBankmRNA BI648460.1 - 2, 3 141 Chr. 17 - 34951231 - 34952457 [-] on Build GRCm38
          GenBankmRNA BI692677.1 - 2, 3 134 Chr. 17 - 34950245 - 34952449 [-] on Build GRCm38
          GenBankmRNA AV515921.1 - 2, 3 129 Chr. 17 - 34950250 - 34952439 [-] on Build GRCm38
          GenBankmRNA BY038602.1 - 2, 3 154 Chr. 17 - 34950450 - 34952466 [-] on Build GRCm38
          GenBankmRNA AW987979.1 - 2, 3 132 Chr. 17 - 34950243 - 34952447 [-] on Build GRCm38
          GenBankmRNA BY323051.1 - 2, 3 150 Chr. 17 - 34950697 - 34952465 [-] on Build GRCm38
          GenBankmRNA AV586528.1 - 2, 3 141 Chr. 17 - 34950314 - 34952455 [-] on Build GRCm38
          GenBankmRNA BU924186.1 - 2, 3 135 Chr. 17 - 34950231 - 34952449 [-] on Build GRCm38
          GenBankmRNA CK792311.1 - 2, 3 179 Chr. 17 - 34950241 - 34952457 [-] on Build GRCm38
          GenBankmRNA AV467561.1 - 2, 3 137 Chr. 17 - 34950286 - 34952455 [-] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix