Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 1110038B12RIK SIRNA Assays ›
          Gene Symbol:
          1110038B12Rik
          Gene Name:
          RIKEN cDNA 1110038B12 gene
          Gene Aliases:
          0610009C20Rik, 0910001G04Rik
          Chromosome Location:
          Chr.17: 34950235 - 34952471 on Build GRCm38
          UniGene ID:
          Mm.28895
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n255425

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 1110038B12Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015536.1 3 190
          NR_027943.1 3 187
          GenBank mRNA AK002372.1 - 3 190
          AK003073.1 - 3 188
          AK004150.1 - 3 188
          BC058123.1 - 3 174
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA CB205178.1 - 3 171 Chr. 17 - 34950231 - 34952452 [-] on Build GRCm38
          GenBankmRNA AW987897.1 - 3 166 Chr. 17 - 34950243 - 34952447 [-] on Build GRCm38
          GenBankmRNA AV488693.1 - 3 171 Chr. 17 - 34950268 - 34952455 [-] on Build GRCm38
          GenBankmRNA CB589057.1 - 3 175 Chr. 17 - 34950231 - 34952456 [-] on Build GRCm38
          GenBankmRNA AV506956.1 - 3 151 Chr. 17 - 34950337 - 34952428 [-] on Build GRCm38
          GenBankmRNA CB321306.1 - 174 Chr. - - [-] on Build
          GenBankmRNA CB588222.1 - 192 Chr. - - [-] on Build
          GenBankmRNA CB196482.1 - 3 171 Chr. 17 - 34950231 - 34952454 [-] on Build GRCm38
          GenBankmRNA CF739395.1 - 3 168 Chr. 17 - 34950241 - 34952449 [-] on Build GRCm38
          GenBankmRNA BX527841.1 - 3 140 Chr. 17 - 34950248 - 34952420 [-] on Build GRCm38
          GenBankmRNA CB575170.1 - 172 Chr. - - [-] on Build
          GenBankmRNA BY307160.1 - 3 186 Chr. 17 - 34950294 - 34952467 [-] on Build GRCm38
          GenBankmRNA BY054987.1 - 3 188 Chr. 17 - 34950450 - 34952466 [-] on Build GRCm38
          GenBankmRNA AV505341.1 - 3 164 Chr. 17 - 34950314 - 34952438 [-] on Build GRCm38
          GenBankmRNA CN694693.1 - 3 176 Chr. 17 - 34951446 - 34952457 [-] on Build GRCm38
          GenBankmRNA BE859815.1 - 3 158 Chr. 17 - 34950252 - 34952439 [-] on Build GRCm38
          GenBankmRNA BY702909.1 - 3 190 Chr. 17 - 34950242 - 34952471 [-] on Build GRCm38
          GenBankmRNA AV479153.1 - 3 154 Chr. 17 - 34950250 - 34952423 [-] on Build GRCm38
          GenBankmRNA AV005223.1 - 3 200 Chr. 17 - 34951748 - 34952468 [-] on Build GRCm38
          GenBankmRNA W66707.1 - 140 Chr. - - [-] on Build
          GenBankmRNA BY086580.1 - 3 184 Chr. 17 - 34950288 - 34952465 [-] on Build GRCm38
          GenBankmRNA BI648460.1 - 3 175 Chr. 17 - 34951231 - 34952457 [-] on Build GRCm38
          GenBankmRNA BI692677.1 - 3 168 Chr. 17 - 34950245 - 34952449 [-] on Build GRCm38
          GenBankmRNA AV515921.1 - 3 163 Chr. 17 - 34950250 - 34952439 [-] on Build GRCm38
          GenBankmRNA BY038602.1 - 3 188 Chr. 17 - 34950450 - 34952466 [-] on Build GRCm38
          GenBankmRNA AW987979.1 - 3 166 Chr. 17 - 34950243 - 34952447 [-] on Build GRCm38
          GenBankmRNA BY323051.1 - 3 184 Chr. 17 - 34950697 - 34952465 [-] on Build GRCm38
          GenBankmRNA AV586528.1 - 3 175 Chr. 17 - 34950314 - 34952455 [-] on Build GRCm38
          GenBankmRNA BU924186.1 - 3 169 Chr. 17 - 34950231 - 34952449 [-] on Build GRCm38
          GenBankmRNA CK792311.1 - 3 213 Chr. 17 - 34950241 - 34952457 [-] on Build GRCm38
          GenBankmRNA AV467561.1 - 3 171 Chr. 17 - 34950286 - 34952455 [-] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0