Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 2410006H16RIK SIRNA Assays ›
          Gene Symbol:
          2410006H16Rik
          Gene Name:
          RIKEN cDNA 2410006H16 gene
          Gene Aliases:
          -
          Chromosome Location:
          Chr.11: 62602877 - 62604806 on Build GRCm38
          UniGene ID:
          Mm.270874
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n256873

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 2410006H16Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_030738.1 1 43
          GenBank mRNA AK010427.1 - 1 44
          AK157519.1 - 1 44
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA W62980.1 - 46 Chr. - - [-] on Build
          GenBankmRNA AA038686.1 - 1 52 Chr. 11 - 62602891 - 62603254 [+] on Build GRCm38
          GenBankmRNA CA469348.1 - 80 Chr. - - [-] on Build
          GenBankmRNA CK795379.1 - 1 33 Chr. 11 - 62602901 - 62604809 [+] on Build GRCm38
          GenBankmRNA AV478545.1 - 1 19 Chr. 11 - 62602901 - 62604804 [+] on Build GRCm38
          GenBankmRNA AV585715.1 - 1 18 Chr. 11 - 62602902 - 62603089 [+] on Build GRCm38
          GenBankmRNA AA087133.1 - 1 17 Chr. 11 - 62602903 - 62604801 [+] on Build GRCm38
          GenBankmRNA CA490873.1 - 80 Chr. - - [-] on Build
          GenBankmRNA DV066609.1 - 66 Chr. - - [-] on Build
          GenBankmRNA W30208.1 - 24 Chr. - - [-] on Build
          GenBankmRNA AV576844.1 - 1 18 Chr. 11 - 62602902 - 62604804 [+] on Build GRCm38
          GenBankmRNA AV471541.1 - 1 21 Chr. 11 - 62602899 - 62603224 [+] on Build GRCm38
          GenBankmRNA AA644831.1 - 1 33 Chr. 11 - 62602896 - 62603637 [+] on Build GRCm38
          GenBankmRNA BY471947.1 - 1 44 Chr. 11 - 62602877 - 62604801 [+] on Build GRCm38
          GenBankmRNA BY123523.1 - 1 40 Chr. 11 - 62602880 - 62604681 [+] on Build GRCm38
          GenBankmRNA AV459516.1 - 1 31 Chr. 11 - 62602892 - 62604809 [+] on Build GRCm38
          GenBankmRNA BY118814.1 - 1 40 Chr. 11 - 62602880 - 62604673 [+] on Build GRCm38
          GenBankmRNA W34684.1 - 18 Chr. - - [-] on Build
          GenBankmRNA CJ176622.1 - 1 40 Chr. 11 - 62602880 - 62604778 [+] on Build GRCm38
          GenBankmRNA BY332380.1 - 1 40 Chr. 11 - 62602880 - 62604672 [+] on Build GRCm38
          GenBankmRNA W29476.1 - 17 Chr. - - [-] on Build
          GenBankmRNA BB652577.1 - 1 40 Chr. 11 - 62602880 - 62603512 [+] on Build GRCm38
          GenBankmRNA BY048877.1 - 1 40 Chr. 11 - 62602880 - 62604470 [+] on Build GRCm38
          GenBankmRNA AV491046.1 - 1 32 Chr. 11 - 62602890 - 62604777 [+] on Build GRCm38
          GenBankmRNA AI060452.1 - 1 30 Chr. 11 - 62602891 - 62604782 [+] on Build GRCm38
          GenBankmRNA AV508300.1 - 1 27 Chr. 11 - 62602893 - 62604804 [+] on Build GRCm38
          GenBankmRNA BY056642.1 - 1 40 Chr. 11 - 62602880 - 62604467 [+] on Build GRCm38
          GenBankmRNA BY709861.1 - 1 39 Chr. 11 - 62602884 - 62604806 [+] on Build GRCm38
          GenBankmRNA DV054738.1 - 64 Chr. - - [-] on Build
          GenBankmRNA AV457388.1 - 1 37 Chr. 11 - 62602883 - 62604778 [+] on Build GRCm38
          GenBankmRNA W13601.1 - 1 16 Chr. 11 - 62602904 - 62604767 [+] on Build GRCm38
          GenBankmRNA AA038258.1 - 1 18 Chr. 11 - 62602902 - 62604457 [+] on Build GRCm38
          GenBankmRNA BE138252.1 - 1 36 Chr. 11 - 62602895 - 62604782 [+] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7b745768ff-vzrjf:80/100.66.128.131:80.
          git-commit: 4b95038805a43672018eef8c57cc3f15d8b0c074
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: integration/2.33.0-meta-desc