Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS4L SIRNA Assays ›
          Gene Symbol:
          Rps4l
          Gene Name:
          ribosomal protein S4-like
          Gene Aliases:
          1110033J19Rik, AA407494, Rps4y2
          Chromosome Location:
          Chr.6: 148354656 - 148355596 on Build GRCm38
          UniGene ID:
          Mm.391542
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n257517

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps4l
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003634.2 1 445
          GenBank mRNA AK004068.1 - 1 446
          AK012382.1 - 1 445
          AK132160.1 - 1 446
          BC047994.1 - 1 417
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AV578314.1 - 1 399 Chr. 6 - 148354702 - 148355170 [+] on Build GRCm38
          GenBankmRNA CK627377.1 - 1 372 Chr. 6 - 148354729 - 148355367 [+] on Build GRCm38
          GenBankmRNA CB204246.1 - 1 414 Chr. 6 - 148354687 - 148355578 [+] on Build GRCm38
          GenBankmRNA AV463957.1 - 1 417 Chr. 6 - 148354683 - 148355197 [+] on Build GRCm38
          GenBankmRNA AV471013.1 - 1 422 Chr. 6 - 148354679 - 148355177 [+] on Build GRCm38
          GenBankmRNA AV516577.1 - 1 399 Chr. 6 - 148354706 - 148355172 [+] on Build GRCm38
          GenBankmRNA AV507738.1 - 1 435 Chr. 6 - 148354667 - 148355175 [+] on Build GRCm38
          GenBankmRNA AV448401.1 - 1 375 Chr. 6 - 148354726 - 148355221 [+] on Build GRCm38
          GenBankmRNA AV514849.1 - 1 435 Chr. 6 - 148354667 - 148355173 [+] on Build GRCm38
          GenBankmRNA CK022865.1 - 450 Chr. - - [-] on Build
          GenBankmRNA CK137553.1 - 1 432 Chr. 6 - 148354669 - 148355260 [+] on Build GRCm38
          GenBankmRNA BG100415.1 - 1 363 Chr. 6 - 148354738 - 148355409 [+] on Build GRCm38
          GenBankmRNA BX516077.1 - 1 430 Chr. 6 - 148354672 - 148355178 [+] on Build GRCm38
          GenBankmRNA AV577061.1 - 1 432 Chr. 6 - 148354669 - 148355212 [+] on Build GRCm38
          GenBankmRNA AV472928.1 - 1 400 Chr. 6 - 148354701 - 148355113 [+] on Build GRCm38
          GenBankmRNA AV469257.1 - 1 387 Chr. 6 - 148354714 - 148355201 [+] on Build GRCm38
          GenBankmRNA AV569749.1 - 1 398 Chr. 6 - 148354709 - 148355277 [+] on Build GRCm38
          GenBankmRNA AA041632.1 - 1 393 Chr. 6 - 148354707 - 148355194 [+] on Build GRCm38
          GenBankmRNA CB182173.1 - 1 441 Chr. 6 - 148354686 - 148355435 [+] on Build GRCm38
          GenBankmRNA AV493656.1 - 1 430 Chr. 6 - 148354670 - 148355111 [+] on Build GRCm38
          GenBankmRNA AV569452.1 - 1 421 Chr. 6 - 148354690 - 148355227 [+] on Build GRCm38
          GenBankmRNA AV453406.1 - 1 432 Chr. 6 - 148354669 - 148355152 [+] on Build GRCm38
          GenBankmRNA AV446887.1 - 1 435 Chr. 6 - 148354667 - 148355149 [+] on Build GRCm38
          GenBankmRNA BU841056.1 - 1 416 Chr. 6 - 148354685 - 148355589 [+] on Build GRCm38
          GenBankmRNA AV583923.1 - 1 375 Chr. 6 - 148354726 - 148355242 [+] on Build GRCm38
          GenBankmRNA AV508759.1 - 1 426 Chr. 6 - 148354675 - 148355151 [+] on Build GRCm38
          GenBankmRNA AV587336.1 - 1 428 Chr. 6 - 148354672 - 148355188 [+] on Build GRCm38
          GenBankmRNA AV583728.1 - 1 426 Chr. 6 - 148354675 - 148355151 [+] on Build GRCm38
          GenBankmRNA BI525490.1 - 1 399 Chr. 6 - 148354709 - 148355592 [+] on Build GRCm38
          GenBankmRNA BU562610.1 - 1 429 Chr. 6 - 148354672 - 148355453 [+] on Build GRCm38
          GenBankmRNA AA269378.1 - 1 360 Chr. 6 - 148354741 - 148355244 [+] on Build GRCm38
          GenBankmRNA AV475819.1 - 1 437 Chr. 6 - 148354673 - 148355149 [+] on Build GRCm38
          GenBankmRNA CB840729.1 - 1 426 Chr. 6 - 148354685 - 148355342 [+] on Build GRCm38
          GenBankmRNA AV452155.1 - 1 396 Chr. 6 - 148354705 - 148355157 [+] on Build GRCm38
          GenBankmRNA CK020541.1 - 1 417 Chr. 6 - 148354684 - 148355385 [+] on Build GRCm38
          GenBankmRNA AV481267.1 - 1 430 Chr. 6 - 148354670 - 148355172 [+] on Build GRCm38
          GenBankmRNA BU551583.1 - 1 394 Chr. 6 - 148354708 - 148355277 [+] on Build GRCm38
          GenBankmRNA AV472068.1 - 1 427 Chr. 6 - 148354680 - 148355149 [+] on Build GRCm38
          GenBankmRNA AV572982.1 - 1 428 Chr. 6 - 148354674 - 148355113 [+] on Build GRCm38
          GenBankmRNA CJ041563.1 - 1 447 Chr. 6 - 148354660 - 148355242 [+] on Build GRCm38
          GenBankmRNA CO797874.1 - 407 Chr. - - [-] on Build
          GenBankmRNA AV587493.1 - 1 387 Chr. 6 - 148354723 - 148355138 [+] on Build GRCm38
          GenBankmRNA AV443982.1 - 1 422 Chr. 6 - 148354682 - 148355170 [+] on Build GRCm38
          GenBankmRNA AV475406.1 - 1 401 Chr. 6 - 148354700 - 148355189 [+] on Build GRCm38
          GenBankmRNA BU898844.1 - 1 429 Chr. 6 - 148354672 - 148355253 [+] on Build GRCm38
          GenBankmRNA AV494619.1 - 1 433 Chr. 6 - 148354668 - 148355181 [+] on Build GRCm38
          GenBankmRNA AW701596.1 - 1 367 Chr. 6 - 148354734 - 148355262 [+] on Build GRCm38
          GenBankmRNA CK031884.1 - 1 409 Chr. 6 - 148354692 - 148355436 [+] on Build GRCm38
          GenBankmRNA CA321842.1 - 1 394 Chr. 6 - 148354706 - 148355409 [+] on Build GRCm38
          GenBankmRNA AV466506.1 - 1 395 Chr. 6 - 148354706 - 148355142 [+] on Build GRCm38
          GenBankmRNA AA000082.1 - 1 438 Chr. 6 - 148354672 - 148355194 [+] on Build GRCm38
          GenBankmRNA AV511109.1 - 1 433 Chr. 6 - 148354668 - 148355135 [+] on Build GRCm38
          GenBankmRNA AV494722.1 - 1 432 Chr. 6 - 148354668 - 148355191 [+] on Build GRCm38
          GenBankmRNA AV474962.1 - 1 428 Chr. 6 - 148354682 - 148355157 [+] on Build GRCm38
          GenBankmRNA AV463010.1 - 1 425 Chr. 6 - 148354674 - 148355148 [+] on Build GRCm38
          GenBankmRNA CB274693.1 - 2 396 Chr. 6 - 148354671 - 148355123 [+] on Build GRCm38
          GenBankmRNA AV513781.1 - 1 386 Chr. 6 - 148354713 - 148355151 [+] on Build GRCm38
          GenBankmRNA AV459279.1 - 1 431 Chr. 6 - 148354670 - 148355154 [+] on Build GRCm38
          GenBankmRNA AV481595.1 - 1 424 Chr. 6 - 148354677 - 148355195 [+] on Build GRCm38
          GenBankmRNA AV464639.1 - 1 417 Chr. 6 - 148354683 - 148355133 [+] on Build GRCm38
          GenBankmRNA CK021457.1 - 1 412 Chr. 6 - 148354688 - 148355532 [+] on Build GRCm38
          GenBankmRNA AV484927.1 - 1 421 Chr. 6 - 148354678 - 148355189 [+] on Build GRCm38
          GenBankmRNA AV573175.1 - 1 423 Chr. 6 - 148354685 - 148355149 [+] on Build GRCm38
          GenBankmRNA AV493615.1 - 1 391 Chr. 6 - 148354712 - 148355196 [+] on Build GRCm38
          GenBankmRNA AV486209.1 - 1 436 Chr. 6 - 148354667 - 148355154 [+] on Build GRCm38
          GenBankmRNA AV488831.1 - 1 401 Chr. 6 - 148354705 - 148355197 [+] on Build GRCm38
          GenBankmRNA AV451783.1 - 1 426 Chr. 6 - 148354674 - 148355143 [+] on Build GRCm38
          GenBankmRNA CJ041148.1 - 1 401 Chr. 6 - 148354703 - 148355150 [+] on Build GRCm38
          GenBankmRNA AV573487.1 - 1 417 Chr. 6 - 148354684 - 148355133 [+] on Build GRCm38
          GenBankmRNA AV586344.1 - 1 391 Chr. 6 - 148354709 - 148355136 [+] on Build GRCm38
          GenBankmRNA AV480251.1 - 1 431 Chr. 6 - 148354685 - 148355145 [+] on Build GRCm38
          GenBankmRNA AV461169.1 - 1 424 Chr. 6 - 148354685 - 148355133 [+] on Build GRCm38
          GenBankmRNA C89050.1 - 446 Chr. - - [-] on Build
          GenBankmRNA AV455874.1 - 1 433 Chr. 6 - 148354668 - 148355142 [+] on Build GRCm38
          GenBankmRNA AV585951.1 - 1 365 Chr. 6 - 148354735 - 148355200 [+] on Build GRCm38
          GenBankmRNA DV662648.1 - 1 434 Chr. 6 - 148354667 - 148355422 [+] on Build GRCm38
          GenBankmRNA CB845206.2 - 1 444 Chr. 6 - 148354667 - 148355453 [+] on Build GRCm38
          GenBankmRNA CB183113.1 - 1 422 Chr. 6 - 148354679 - 148355475 [+] on Build GRCm38
          GenBankmRNA CK031938.1 - 1 409 Chr. 6 - 148354692 - 148355385 [+] on Build GRCm38
          GenBankmRNA AV491120.1 - 1 424 Chr. 6 - 148354677 - 148355193 [+] on Build GRCm38
          GenBankmRNA AV470717.1 - 1 402 Chr. 6 - 148354703 - 148355151 [+] on Build GRCm38
          GenBankmRNA AV485052.1 - 1 417 Chr. 6 - 148354684 - 148355198 [+] on Build GRCm38
          GenBankmRNA AV490188.1 - 1 437 Chr. 6 - 148354667 - 148355176 [+] on Build GRCm38
          GenBankmRNA AV509662.1 - 1 430 Chr. 6 - 148354669 - 148355152 [+] on Build GRCm38
          GenBankmRNA AV585136.1 - 1 423 Chr. 6 - 148354678 - 148355194 [+] on Build GRCm38
          GenBankmRNA AV444821.1 - 1 423 Chr. 6 - 148354691 - 148355191 [+] on Build GRCm38
          GenBankmRNA AV455637.1 - 1 422 Chr. 6 - 148354690 - 148355179 [+] on Build GRCm38
          GenBankmRNA AV457756.1 - 1 426 Chr. 6 - 148354673 - 148355144 [+] on Build GRCm38
          GenBankmRNA AV496022.1 - 1 417 Chr. 6 - 148354684 - 148355180 [+] on Build GRCm38
          GenBankmRNA BI904197.1 - 1 388 Chr. 6 - 148354749 - 148355474 [+] on Build GRCm38
          GenBankmRNA AV493657.1 - 1 428 Chr. 6 - 148354672 - 148355152 [+] on Build GRCm38
          GenBankmRNA AV491618.1 - 1 412 Chr. 6 - 148354702 - 148355197 [+] on Build GRCm38
          GenBankmRNA AV499469.1 - 1 432 Chr. 6 - 148354671 - 148355173 [+] on Build GRCm38
          GenBankmRNA AV573947.1 - 1 416 Chr. 6 - 148354685 - 148355142 [+] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix