Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RPS4L SIRNA Assays ›
          Gene Symbol:
          Rps4l
          Gene Name:
          ribosomal protein S4-like
          Gene Aliases:
          1110033J19Rik, AA407494, Rps4y2
          Chromosome Location:
          Chr.6: 148354656 - 148355596 on Build GRCm38
          UniGene ID:
          Mm.391542
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID n257522

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rps4l
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_003634.2 1 112
          GenBank mRNA AK004068.1 - 1 113
          AK012382.1 - 1 113
          AK132160.1 - 1 113
          BC047994.1 - 1 84
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA AV511384.1 - 1 96 Chr. 6 - 148354670 - 148355107 [+] on Build GRCm38
          GenBankmRNA AV578314.1 - 1 66 Chr. 6 - 148354702 - 148355170 [+] on Build GRCm38
          GenBankmRNA CK627377.1 - 1 39 Chr. 6 - 148354729 - 148355367 [+] on Build GRCm38
          GenBankmRNA CB204246.1 - 1 81 Chr. 6 - 148354687 - 148355578 [+] on Build GRCm38
          GenBankmRNA AV463957.1 - 1 84 Chr. 6 - 148354683 - 148355197 [+] on Build GRCm38
          GenBankmRNA AV507738.1 - 1 102 Chr. 6 - 148354667 - 148355175 [+] on Build GRCm38
          GenBankmRNA AV471013.1 - 1 89 Chr. 6 - 148354679 - 148355177 [+] on Build GRCm38
          GenBankmRNA AV448401.1 - 1 42 Chr. 6 - 148354726 - 148355221 [+] on Build GRCm38
          GenBankmRNA BY328788.1 - 1 110 Chr. 6 - 148354658 - 148354990 [+] on Build GRCm38
          GenBankmRNA CJ163072.1 - 1 110 Chr. 6 - 148354658 - 148355047 [+] on Build GRCm38
          GenBankmRNA AV514849.1 - 1 102 Chr. 6 - 148354667 - 148355173 [+] on Build GRCm38
          GenBankmRNA CK022865.1 - 117 Chr. - - [-] on Build
          GenBankmRNA AV462502.1 - 1 92 Chr. 6 - 148354677 - 148355103 [+] on Build GRCm38
          GenBankmRNA CK137553.1 - 1 99 Chr. 6 - 148354669 - 148355260 [+] on Build GRCm38
          GenBankmRNA BG100415.1 - 1 30 Chr. 6 - 148354738 - 148355409 [+] on Build GRCm38
          GenBankmRNA BX516077.1 - 1 97 Chr. 6 - 148354672 - 148355178 [+] on Build GRCm38
          GenBankmRNA AV511272.1 - 1 100 Chr. 6 - 148354667 - 148355095 [+] on Build GRCm38
          GenBankmRNA AV577061.1 - 1 99 Chr. 6 - 148354669 - 148355212 [+] on Build GRCm38
          GenBankmRNA AV475843.1 - 1 98 Chr. 6 - 148354668 - 148354978 [+] on Build GRCm38
          GenBankmRNA BY114163.1 - 1 110 Chr. 6 - 148354658 - 148354973 [+] on Build GRCm38
          GenBankmRNA AV472928.1 - 1 67 Chr. 6 - 148354701 - 148355113 [+] on Build GRCm38
          GenBankmRNA AV469257.1 - 1 54 Chr. 6 - 148354714 - 148355201 [+] on Build GRCm38
          GenBankmRNA AV463810.1 - 1 102 Chr. 6 - 148354667 - 148355103 [+] on Build GRCm38
          GenBankmRNA BY115419.1 - 1 110 Chr. 6 - 148354658 - 148354996 [+] on Build GRCm38
          GenBankmRNA AA007834.1 - 1 97 Chr. 6 - 148354671 - 148355250 [+] on Build GRCm38
          GenBankmRNA AA041632.1 - 1 61 Chr. 6 - 148354707 - 148355194 [+] on Build GRCm38
          GenBankmRNA AV516234.1 - 1 81 Chr. 6 - 148354691 - 148355068 [+] on Build GRCm38
          GenBankmRNA CB182173.1 - 1 108 Chr. 6 - 148354686 - 148355435 [+] on Build GRCm38
          GenBankmRNA AV493656.1 - 1 97 Chr. 6 - 148354670 - 148355111 [+] on Build GRCm38
          GenBankmRNA AV483695.1 - 1 91 Chr. 6 - 148354682 - 148355097 [+] on Build GRCm38
          GenBankmRNA AV569452.1 - 1 88 Chr. 6 - 148354690 - 148355227 [+] on Build GRCm38
          GenBankmRNA AV453406.1 - 1 99 Chr. 6 - 148354669 - 148355152 [+] on Build GRCm38
          GenBankmRNA AV446887.1 - 1 102 Chr. 6 - 148354667 - 148355149 [+] on Build GRCm38
          GenBankmRNA AV472654.1 - 1 87 Chr. 6 - 148354681 - 148355143 [+] on Build GRCm38
          GenBankmRNA BU841056.1 - 1 83 Chr. 6 - 148354685 - 148355589 [+] on Build GRCm38
          GenBankmRNA AV583923.1 - 1 42 Chr. 6 - 148354726 - 148355242 [+] on Build GRCm38
          GenBankmRNA AV508759.1 - 1 93 Chr. 6 - 148354675 - 148355151 [+] on Build GRCm38
          GenBankmRNA AV587336.1 - 1 95 Chr. 6 - 148354672 - 148355188 [+] on Build GRCm38
          GenBankmRNA AV583728.1 - 1 93 Chr. 6 - 148354675 - 148355151 [+] on Build GRCm38
          GenBankmRNA CJ171908.1 - 1 82 Chr. 6 - 148354687 - 148355065 [+] on Build GRCm38
          GenBankmRNA BI525490.1 - 1 66 Chr. 6 - 148354709 - 148355592 [+] on Build GRCm38
          GenBankmRNA BU562610.1 - 1 96 Chr. 6 - 148354672 - 148355453 [+] on Build GRCm38
          GenBankmRNA AV496116.1 - 1 96 Chr. 6 - 148354672 - 148355008 [+] on Build GRCm38
          GenBankmRNA AV515777.1 - 1 102 Chr. 6 - 148354667 - 148355103 [+] on Build GRCm38
          GenBankmRNA AV506963.1 - 1 93 Chr. 6 - 148354677 - 148355017 [+] on Build GRCm38
          GenBankmRNA AA269378.1 - 1 27 Chr. 6 - 148354741 - 148355244 [+] on Build GRCm38
          GenBankmRNA AV475819.1 - 1 103 Chr. 6 - 148354673 - 148355149 [+] on Build GRCm38
          GenBankmRNA CB840729.1 - 1 93 Chr. 6 - 148354685 - 148355342 [+] on Build GRCm38
          GenBankmRNA AV452155.1 - 1 63 Chr. 6 - 148354705 - 148355157 [+] on Build GRCm38
          GenBankmRNA CK020541.1 - 1 84 Chr. 6 - 148354684 - 148355385 [+] on Build GRCm38
          GenBankmRNA AV506996.1 - 1 95 Chr. 6 - 148354682 - 148355099 [+] on Build GRCm38
          GenBankmRNA AV481267.1 - 1 97 Chr. 6 - 148354670 - 148355172 [+] on Build GRCm38
          GenBankmRNA AV467263.1 - 1 83 Chr. 6 - 148354684 - 148355103 [+] on Build GRCm38
          GenBankmRNA BU551583.1 - 1 60 Chr. 6 - 148354708 - 148355277 [+] on Build GRCm38
          GenBankmRNA AV472068.1 - 1 94 Chr. 6 - 148354680 - 148355149 [+] on Build GRCm38
          GenBankmRNA AV572982.1 - 1 95 Chr. 6 - 148354674 - 148355113 [+] on Build GRCm38
          GenBankmRNA CJ041563.1 - 1 113 Chr. 6 - 148354660 - 148355242 [+] on Build GRCm38
          GenBankmRNA CO797874.1 - 74 Chr. - - [-] on Build
          GenBankmRNA BY321323.1 - 1 58 Chr. 6 - 148354710 - 148355056 [+] on Build GRCm38
          GenBankmRNA AV472933.1 - 1 89 Chr. 6 - 148354685 - 148355069 [+] on Build GRCm38
          GenBankmRNA AV443982.1 - 1 89 Chr. 6 - 148354682 - 148355170 [+] on Build GRCm38
          GenBankmRNA AV587493.1 - 1 54 Chr. 6 - 148354723 - 148355138 [+] on Build GRCm38
          GenBankmRNA AI325738.1 - 1 47 Chr. 6 - 148354721 - 148355098 [+] on Build GRCm38
          GenBankmRNA AV475406.1 - 1 68 Chr. 6 - 148354700 - 148355189 [+] on Build GRCm38
          GenBankmRNA AV494619.1 - 1 100 Chr. 6 - 148354668 - 148355181 [+] on Build GRCm38
          GenBankmRNA BU898844.1 - 1 96 Chr. 6 - 148354672 - 148355253 [+] on Build GRCm38
          GenBankmRNA CK031884.1 - 1 76 Chr. 6 - 148354692 - 148355436 [+] on Build GRCm38
          GenBankmRNA AW701596.1 - 1 34 Chr. 6 - 148354734 - 148355262 [+] on Build GRCm38
          GenBankmRNA CA321842.1 - 1 61 Chr. 6 - 148354706 - 148355409 [+] on Build GRCm38
          GenBankmRNA AV466506.1 - 1 62 Chr. 6 - 148354706 - 148355142 [+] on Build GRCm38
          GenBankmRNA AA000082.1 - 1 106 Chr. 6 - 148354672 - 148355194 [+] on Build GRCm38
          GenBankmRNA BF153001.1 - 1 118 Chr. 6 - 148354672 - 148355081 [+] on Build GRCm38
          GenBankmRNA AV510526.1 - 1 98 Chr. 6 - 148354667 - 148355092 [+] on Build GRCm38
          GenBankmRNA AV498101.1 - 1 105 Chr. 6 - 148354662 - 148355099 [+] on Build GRCm38
          GenBankmRNA AV584069.1 - 1 95 Chr. 6 - 148354677 - 148355061 [+] on Build GRCm38
          GenBankmRNA AV482075.1 - 1 87 Chr. 6 - 148354682 - 148355090 [+] on Build GRCm38
          GenBankmRNA AV511109.1 - 1 100 Chr. 6 - 148354668 - 148355135 [+] on Build GRCm38
          GenBankmRNA AV494722.1 - 1 99 Chr. 6 - 148354668 - 148355191 [+] on Build GRCm38
          GenBankmRNA AV474962.1 - 1 95 Chr. 6 - 148354682 - 148355157 [+] on Build GRCm38
          GenBankmRNA AV463010.1 - 1 92 Chr. 6 - 148354674 - 148355148 [+] on Build GRCm38
          GenBankmRNA CJ164673.1 - 1 110 Chr. 6 - 148354658 - 148355078 [+] on Build GRCm38
          GenBankmRNA CB274693.1 - 1, 2 63 Chr. 6 - 148354671 - 148355123 [+] on Build GRCm38
          GenBankmRNA AV451715.1 - 1 98 Chr. 6 - 148354669 - 148355098 [+] on Build GRCm38
          GenBankmRNA AV513781.1 - 1 53 Chr. 6 - 148354713 - 148355151 [+] on Build GRCm38
          GenBankmRNA AA028728.1 - 33 Chr. - - [-] on Build
          GenBankmRNA AV459279.1 - 1 98 Chr. 6 - 148354670 - 148355154 [+] on Build GRCm38
          GenBankmRNA AV581989.1 - 1 90 Chr. 6 - 148354678 - 148354982 [+] on Build GRCm38
          GenBankmRNA AV481595.1 - 1 91 Chr. 6 - 148354677 - 148355195 [+] on Build GRCm38
          GenBankmRNA CK021457.1 - 1 79 Chr. 6 - 148354688 - 148355532 [+] on Build GRCm38
          GenBankmRNA AV484927.1 - 1 88 Chr. 6 - 148354678 - 148355189 [+] on Build GRCm38
          GenBankmRNA CJ040911.1 - 1 111 Chr. 6 - 148354658 - 148355273 [+] on Build GRCm38
          GenBankmRNA AV573175.1 - 1 90 Chr. 6 - 148354685 - 148355149 [+] on Build GRCm38
          GenBankmRNA AL117823.1 - 1 106 Chr. 6 - 148354667 - 148354928 [+] on Build GRCm38
          GenBankmRNA AV493615.1 - 1 58 Chr. 6 - 148354712 - 148355196 [+] on Build GRCm38
          GenBankmRNA BF012952.1 - 1 33 Chr. 6 - 148354735 - 148355078 [+] on Build GRCm38
          GenBankmRNA AV486209.1 - 1 103 Chr. 6 - 148354667 - 148355154 [+] on Build GRCm38
          GenBankmRNA AV488831.1 - 1 68 Chr. 6 - 148354705 - 148355197 [+] on Build GRCm38
          GenBankmRNA AV451783.1 - 1 93 Chr. 6 - 148354674 - 148355143 [+] on Build GRCm38
          GenBankmRNA BY096065.1 - 1 89 Chr. 6 - 148354680 - 148355071 [+] on Build GRCm38
          GenBankmRNA AV493432.1 - 1 70 Chr. 6 - 148354698 - 148355087 [+] on Build GRCm38
          GenBankmRNA CJ041148.1 - 1 68 Chr. 6 - 148354703 - 148355150 [+] on Build GRCm38
          GenBankmRNA AV573487.1 - 1 84 Chr. 6 - 148354684 - 148355133 [+] on Build GRCm38
          GenBankmRNA AV586344.1 - 1 58 Chr. 6 - 148354709 - 148355136 [+] on Build GRCm38
          GenBankmRNA AV480251.1 - 1 98 Chr. 6 - 148354685 - 148355145 [+] on Build GRCm38
          GenBankmRNA AV461169.1 - 1 91 Chr. 6 - 148354685 - 148355133 [+] on Build GRCm38
          GenBankmRNA C89050.1 - 113 Chr. - - [-] on Build
          GenBankmRNA AV465962.1 - 1 100 Chr. 6 - 148354667 - 148354985 [+] on Build GRCm38
          GenBankmRNA AV506464.1 - 1 93 Chr. 6 - 148354685 - 148355100 [+] on Build GRCm38
          GenBankmRNA AV455874.1 - 1 100 Chr. 6 - 148354668 - 148355142 [+] on Build GRCm38
          GenBankmRNA AV503345.1 - 1 89 Chr. 6 - 148354682 - 148355096 [+] on Build GRCm38
          GenBankmRNA AV495870.1 - 1 97 Chr. 6 - 148354672 - 148355169 [+] on Build GRCm38
          GenBankmRNA DV662648.1 - 1 101 Chr. 6 - 148354667 - 148355422 [+] on Build GRCm38
          GenBankmRNA CB845206.2 - 1 111 Chr. 6 - 148354667 - 148355453 [+] on Build GRCm38
          GenBankmRNA CB183113.1 - 1 89 Chr. 6 - 148354679 - 148355475 [+] on Build GRCm38
          GenBankmRNA CK031938.1 - 1 76 Chr. 6 - 148354692 - 148355385 [+] on Build GRCm38
          GenBankmRNA AV491120.1 - 1 91 Chr. 6 - 148354677 - 148355193 [+] on Build GRCm38
          GenBankmRNA AV470717.1 - 1 69 Chr. 6 - 148354703 - 148355151 [+] on Build GRCm38
          GenBankmRNA AV485052.1 - 1 84 Chr. 6 - 148354684 - 148355198 [+] on Build GRCm38
          GenBankmRNA AV490188.1 - 1 104 Chr. 6 - 148354667 - 148355176 [+] on Build GRCm38
          GenBankmRNA AV509662.1 - 1 97 Chr. 6 - 148354669 - 148355152 [+] on Build GRCm38
          GenBankmRNA AV444821.1 - 1 90 Chr. 6 - 148354691 - 148355191 [+] on Build GRCm38
          GenBankmRNA AV585136.1 - 1 90 Chr. 6 - 148354678 - 148355194 [+] on Build GRCm38
          GenBankmRNA AV455637.1 - 1 89 Chr. 6 - 148354690 - 148355179 [+] on Build GRCm38
          GenBankmRNA AV457756.1 - 1 93 Chr. 6 - 148354673 - 148355144 [+] on Build GRCm38
          GenBankmRNA AV496022.1 - 1 84 Chr. 6 - 148354684 - 148355180 [+] on Build GRCm38
          GenBankmRNA BI904197.1 - 1 55 Chr. 6 - 148354749 - 148355474 [+] on Build GRCm38
          GenBankmRNA BY034424.1 - 1 58 Chr. 6 - 148354710 - 148355085 [+] on Build GRCm38
          GenBankmRNA AA073401.1 - 1 47 Chr. 6 - 148354721 - 148355163 [+] on Build GRCm38
          GenBankmRNA AV493657.1 - 1 95 Chr. 6 - 148354672 - 148355152 [+] on Build GRCm38
          GenBankmRNA AV491618.1 - 1 79 Chr. 6 - 148354702 - 148355197 [+] on Build GRCm38
          GenBankmRNA AV499469.1 - 1 99 Chr. 6 - 148354671 - 148355173 [+] on Build GRCm38
          GenBankmRNA AV573947.1 - 1 83 Chr. 6 - 148354685 - 148355142 [+] on Build GRCm38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          biological_process
          molecular_function

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix