Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp672 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_028331.2 | 1, 2 | 100 | |||
NR_046280.1 | 1, 2 | 100 | ||||
GenBank mRNA | AK008471.1 | - | 1, 2 | 76 | ||
AK019676.1 | - | 1, 2 | 50 | |||
AK085166.1 | - | 1, 2 | 60 | |||
BC034706.1 | - | 1, 2 | 47 | 3582315 | ||
BG091612.1 | - | 1, 2 | 61 | |||
BU148968.1 | - | 1, 2 | 32 | |||
BY017375.1 | - | 1, 2 | 81 | |||
BY715308.1 | - | 1, 2 | 50 | |||
CA321162.1 | - | 1, 2 | 33 | |||
CV783882.1 | - | 1, 2 | 49 | |||
DV654279.1 | - | 1, 2 | 134 |