Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EPB41L4A-AS1 SIRNA Assays ›
          Gene Symbol:
          EPB41L4A-AS1
          Gene Name:
          EPB41L4A antisense RNA 1
          Gene Aliases:
          C5orf26, NCRNA00219, TIGA1
          Chromosome Location:
          Chr.5: 112160526 - 112162501 on Build GRCh38
          UniGene ID:
          Hs.12082
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258476

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EPB41L4A-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015370.2 2 808
          GenBank mRNA AB002437.1 - 2 440
          AB048207.1 - 2 808
          DA266578.1 - 2 176
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BG331014.1 - 2 453 Chr. 5 - 112160881 - 112162273 [+] on Build GRCh38
          GenBankmRNA HY107502.1 - 2 468 Chr. 5 - 112160866 - 112162195 [+] on Build GRCh38
          GenBankmRNA DA793818.1 - 2 469 Chr. 5 - 112160865 - 112162192 [+] on Build GRCh38
          GenBankmRNA CK005513.1 - 2 157 Chr. 5 - 112161179 - 112162571 [+] on Build GRCh38
          GenBankmRNA DA072630.1 - 2 459 Chr. 5 - 112160873 - 112162227 [+] on Build GRCh38
          GenBankmRNA BE220690.1 - 2 426 Chr. 5 - 112160908 - 112162168 [+] on Build GRCh38
          GenBankmRNA BP331802.1 - 2 475 Chr. 5 - 112160860 - 112162216 [+] on Build GRCh38
          GenBankmRNA HY089009.1 - 2 467 Chr. 5 - 112160866 - 112161776 [+] on Build GRCh38
          GenBankmRNA BI768890.1 - 2 450 Chr. 5 - 112160880 - 112162382 [+] on Build GRCh38
          GenBankmRNA AV688894.1 - 2 169 Chr. 5 - 112161178 - 112162307 [+] on Build GRCh38
          GenBankmRNA BE786095.1 - 2 431 Chr. 5 - 112160903 - 112162195 [+] on Build GRCh38
          GenBankmRNA DA770717.1 - 2 469 Chr. 5 - 112160865 - 112162235 [+] on Build GRCh38
          GenBankmRNA BM547410.1 - 2 468 Chr. 5 - 112160880 - 112162321 [+] on Build GRCh38
          GenBankmRNA BF969780.1 - 2 168 Chr. 5 - 112161168 - 112162315 [+] on Build GRCh38
          GenBankmRNA BI756062.1 - 2 453 Chr. 5 - 112160880 - 112162306 [+] on Build GRCh38
          GenBankmRNA DA266177.1 - 2 473 Chr. 5 - 112160866 - 112162186 [+] on Build GRCh38
          GenBankmRNA AI342959.1 - 2 403 Chr. 5 - 112160931 - 112161759 [+] on Build GRCh38
          GenBankmRNA CF123850.1 - 2 499 Chr. 5 - 112160873 - 112162258 [+] on Build GRCh38
          GenBankmRNA CF127270.1 - 2 427 Chr. 5 - 112160913 - 112162312 [+] on Build GRCh38
          GenBankmRNA CN301203.1 - 2 511 Chr. 5 - 112160881 - 112162303 [+] on Build GRCh38
          GenBankmRNA DA124384.1 - 2 469 Chr. 5 - 112160865 - 112161774 [+] on Build GRCh38
          GenBankmRNA BF727429.1 - 2 451 Chr. 5 - 112160883 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA532218.1 - 2 470 Chr. 5 - 112160864 - 112162250 [+] on Build GRCh38
          GenBankmRNA DA528195.1 - 2 469 Chr. 5 - 112160865 - 112162250 [+] on Build GRCh38
          GenBankmRNA DA769288.1 - 2 462 Chr. 5 - 112160872 - 112161774 [+] on Build GRCh38
          GenBankmRNA DA787078.1 - 2 442 Chr. 5 - 112160896 - 112161883 [+] on Build GRCh38
          GenBankmRNA AL542607.3 - 2 455 Chr. 5 - 112160880 - 112162294 [+] on Build GRCh38
          GenBankmRNA DA293095.1 - 2 356 Chr. 5 - 112160978 - 112162319 [+] on Build GRCh38
          GenBankmRNA DB269453.1 - 2 470 Chr. 5 - 112160864 - 112162209 [+] on Build GRCh38
          GenBankmRNA DA207067.1 - 2 472 Chr. 5 - 112160866 - 112162219 [+] on Build GRCh38
          GenBankmRNA DA925618.1 - 2 470 Chr. 5 - 112160865 - 112162232 [+] on Build GRCh38
          GenBankmRNA CB155341.1 - 2 469 Chr. 5 - 112160865 - 112161772 [+] on Build GRCh38
          GenBankmRNA BF791660.1 - 448 Chr. - - [-] on Build
          GenBankmRNA HY152231.1 - 2 464 Chr. 5 - 112160870 - 112161760 [+] on Build GRCh38
          GenBankmRNA BE259773.1 - 2 423 Chr. 5 - 112160911 - 112162310 [+] on Build GRCh38
          GenBankmRNA BF446029.1 - 2 456 Chr. 5 - 112160878 - 112161773 [+] on Build GRCh38
          GenBankmRNA HY119139.1 - 2 468 Chr. 5 - 112160866 - 112161774 [+] on Build GRCh38
          GenBankmRNA CD176026.1 - 2 485 Chr. 5 - 112160855 - 112162308 [+] on Build GRCh38
          GenBankmRNA AL551513.3 - 2 483 Chr. 5 - 112160851 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA506254.1 - 2 469 Chr. 5 - 112160865 - 112161774 [+] on Build GRCh38
          GenBankmRNA BE670313.1 - 2 453 Chr. 5 - 112160881 - 112161772 [+] on Build GRCh38
          GenBankmRNA DB171189.1 - 2 469 Chr. 5 - 112160865 - 112162231 [+] on Build GRCh38
          GenBankmRNA DA269274.1 - 2 467 Chr. 5 - 112160865 - 112162206 [+] on Build GRCh38
          GenBankmRNA AW572317.1 - 2 464 Chr. 5 - 112160870 - 112161776 [+] on Build GRCh38
          GenBankmRNA DA305841.1 - 2 469 Chr. 5 - 112160865 - 112162212 [+] on Build GRCh38
          GenBankmRNA AA648113.1 - 2 304 Chr. 5 - 112161035 - 112162287 [+] on Build GRCh38
          GenBankmRNA CN301200.1 - 2 449 Chr. 5 - 112160885 - 112162303 [+] on Build GRCh38
          GenBankmRNA AA496650.1 - 2 319 Chr. 5 - 112161027 - 112162277 [+] on Build GRCh38
          GenBankmRNA DA495234.1 - 2 472 Chr. 5 - 112160866 - 112162226 [+] on Build GRCh38
          GenBankmRNA F36336.1 - 199 Chr. - - [-] on Build
          GenBankmRNA BM786647.1 - 2 459 Chr. 5 - 112160874 - 112162280 [+] on Build GRCh38
          GenBankmRNA DA393479.1 - 2 469 Chr. 5 - 112160865 - 112162239 [+] on Build GRCh38
          GenBankmRNA HY034482.1 - 2 462 Chr. 5 - 112160870 - 112162190 [+] on Build GRCh38
          GenBankmRNA CN301208.1 - 2 446 Chr. 5 - 112160888 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA902367.1 - 2 469 Chr. 5 - 112160865 - 112162229 [+] on Build GRCh38
          GenBankmRNA CD248174.1 - 485 Chr. - - [-] on Build
          GenBankmRNA AU253876.1 - 2 470 Chr. 5 - 112160864 - 112162290 [+] on Build GRCh38
          GenBankmRNA BU170152.1 - 2 447 Chr. 5 - 112160887 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA531926.1 - 2 468 Chr. 5 - 112160865 - 112162239 [+] on Build GRCh38
          GenBankmRNA DA145323.1 - 2 471 Chr. 5 - 112160866 - 112162216 [+] on Build GRCh38
          GenBankmRNA DA179074.1 - 2 472 Chr. 5 - 112160866 - 112162215 [+] on Build GRCh38
          GenBankmRNA DA509900.1 - 2 469 Chr. 5 - 112160865 - 112162213 [+] on Build GRCh38
          GenBankmRNA BM745358.1 - 2 471 Chr. 5 - 112160866 - 112161776 [+] on Build GRCh38
          GenBankmRNA DA747400.1 - 2 470 Chr. 5 - 112160864 - 112162204 [+] on Build GRCh38
          GenBankmRNA DB526340.1 - 2 254 Chr. 5 - 112161080 - 112162306 [+] on Build GRCh38
          GenBankmRNA DA499568.1 - 2 469 Chr. 5 - 112160865 - 112161772 [+] on Build GRCh38
          GenBankmRNA AI879105.1 - 2 460 Chr. 5 - 112160874 - 112161766 [+] on Build GRCh38
          GenBankmRNA DA519930.1 - 2 468 Chr. 5 - 112160865 - 112162214 [+] on Build GRCh38
          GenBankmRNA AW954951.1 - 2 430 Chr. 5 - 112160902 - 112162298 [+] on Build GRCh38
          GenBankmRNA DA782287.1 - 2 470 Chr. 5 - 112160864 - 112162225 [+] on Build GRCh38
          GenBankmRNA DA681460.1 - 2 478 Chr. 5 - 112160866 - 112162231 [+] on Build GRCh38
          GenBankmRNA DB532978.1 - 2 254 Chr. 5 - 112161081 - 112162306 [+] on Build GRCh38
          GenBankmRNA DA076661.1 - 2 469 Chr. 5 - 112160864 - 112162221 [+] on Build GRCh38
          GenBankmRNA BF314746.1 - 446 Chr. - - [-] on Build
          GenBankmRNA BM695362.1 - 2 448 Chr. 5 - 112160886 - 112162307 [+] on Build GRCh38
          GenBankmRNA BF967293.2 - 2 474 Chr. 5 - 112160862 - 112162294 [+] on Build GRCh38
          GenBankmRNA BE392562.1 - 2 426 Chr. 5 - 112160905 - 112162305 [+] on Build GRCh38
          GenBankmRNA DA121255.1 - 2 461 Chr. 5 - 112160873 - 112162210 [+] on Build GRCh38
          GenBankmRNA BG385395.1 - 2 477 Chr. 5 - 112160881 - 112161776 [+] on Build GRCh38
          GenBankmRNA DA856584.1 - 2 469 Chr. 5 - 112160865 - 112162209 [+] on Build GRCh38
          GenBankmRNA BM781998.1 - 2 469 Chr. 5 - 112160866 - 112162268 [+] on Build GRCh38
          GenBankmRNA DA083945.1 - 2 467 Chr. 5 - 112160865 - 112162206 [+] on Build GRCh38
          GenBankmRNA DA500844.1 - 2 467 Chr. 5 - 112160867 - 112162209 [+] on Build GRCh38
          GenBankmRNA DA517047.1 - 2 471 Chr. 5 - 112160864 - 112162188 [+] on Build GRCh38
          GenBankmRNA DA502642.1 - 2 471 Chr. 5 - 112160866 - 112162210 [+] on Build GRCh38
          GenBankmRNA BQ272145.1 - 2 447 Chr. 5 - 112160887 - 112162187 [+] on Build GRCh38
          GenBankmRNA DA210552.1 - 2 468 Chr. 5 - 112160866 - 112162212 [+] on Build GRCh38
          GenBankmRNA DA779776.1 - 2 471 Chr. 5 - 112160866 - 112162222 [+] on Build GRCh38
          GenBankmRNA BU196182.1 - 453 Chr. - - [-] on Build
          GenBankmRNA DA745136.1 - 2 440 Chr. 5 - 112160896 - 112162227 [+] on Build GRCh38
          GenBankmRNA DA287168.1 - 2 441 Chr. 5 - 112160894 - 112162249 [+] on Build GRCh38
          GenBankmRNA BE616318.1 - 421 Chr. - - [-] on Build
          GenBankmRNA DA174935.1 - 2 462 Chr. 5 - 112160873 - 112161776 [+] on Build GRCh38
          GenBankmRNA DA522279.1 - 2 472 Chr. 5 - 112160866 - 112162209 [+] on Build GRCh38
          GenBankmRNA BE395909.1 - 2 427 Chr. 5 - 112160905 - 112162306 [+] on Build GRCh38
          GenBankmRNA DA625101.1 - 2 469 Chr. 5 - 112160865 - 112162197 [+] on Build GRCh38
          GenBankmRNA AA248772.1 - 2 143 Chr. 5 - 112161192 - 112162298 [+] on Build GRCh38
          GenBankmRNA BX115634.1 - 2 307 Chr. 5 - 112161027 - 112162315 [+] on Build GRCh38
          GenBankmRNA BF314470.1 - 446 Chr. - - [-] on Build
          GenBankmRNA CB125170.1 - 2 461 Chr. 5 - 112160873 - 112162260 [+] on Build GRCh38
          GenBankmRNA CN301202.1 - 2 433 Chr. 5 - 112160901 - 112162307 [+] on Build GRCh38
          GenBankmRNA HY027741.1 - 2 470 Chr. 5 - 112160866 - 112161776 [+] on Build GRCh38
          GenBankmRNA BP343090.1 - 2 470 Chr. 5 - 112160864 - 112162227 [+] on Build GRCh38
          GenBankmRNA DA907585.1 - 2 467 Chr. 5 - 112160867 - 112162206 [+] on Build GRCh38
          GenBankmRNA DA503766.1 - 2 469 Chr. 5 - 112160865 - 112162202 [+] on Build GRCh38
          GenBankmRNA BF058851.1 - 2 456 Chr. 5 - 112160878 - 112161761 [+] on Build GRCh38
          GenBankmRNA BP358751.1 - 2 470 Chr. 5 - 112160864 - 112162220 [+] on Build GRCh38
          GenBankmRNA CB124040.1 - 2 470 Chr. 5 - 112160864 - 112161762 [+] on Build GRCh38
          GenBankmRNA BE313436.1 - 2 429 Chr. 5 - 112160905 - 112162290 [+] on Build GRCh38
          GenBankmRNA DA116174.1 - 2 476 Chr. 5 - 112160860 - 112162226 [+] on Build GRCh38
          GenBankmRNA AA648234.1 - 2 303 Chr. 5 - 112161035 - 112161758 [+] on Build GRCh38
          GenBankmRNA DA790029.1 - 2 470 Chr. 5 - 112160864 - 112162220 [+] on Build GRCh38
          GenBankmRNA DA260934.1 - 2 469 Chr. 5 - 112160865 - 112162215 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix