Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EPB41L4A-AS1 SIRNA Assays ›
          Gene Symbol:
          EPB41L4A-AS1
          Gene Name:
          EPB41L4A antisense RNA 1
          Gene Aliases:
          C5orf26, NCRNA00219, TIGA1
          Chromosome Location:
          Chr.5: 112160526 - 112162501 on Build GRCh38
          UniGene ID:
          Hs.12082
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n258480

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EPB41L4A-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015370.2 3 891
          GenBank mRNA AB002437.1 - 3 523
          AB048207.1 - 3 891
          DA266578.1 - 3 259
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU170152.1 - 3 530 Chr. 5 - 112160887 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA531926.1 - 3 551 Chr. 5 - 112160865 - 112162239 [+] on Build GRCh38
          GenBankmRNA DA145323.1 - 3 554 Chr. 5 - 112160866 - 112162216 [+] on Build GRCh38
          GenBankmRNA DA179074.1 - 3 555 Chr. 5 - 112160866 - 112162215 [+] on Build GRCh38
          GenBankmRNA DA509900.1 - 3 552 Chr. 5 - 112160865 - 112162213 [+] on Build GRCh38
          GenBankmRNA CK005513.1 - 3 240 Chr. 5 - 112161179 - 112162571 [+] on Build GRCh38
          GenBankmRNA DB526340.1 - 3 337 Chr. 5 - 112161080 - 112162306 [+] on Build GRCh38
          GenBankmRNA DA072630.1 - 3 542 Chr. 5 - 112160873 - 112162227 [+] on Build GRCh38
          GenBankmRNA BP331802.1 - 3 558 Chr. 5 - 112160860 - 112162216 [+] on Build GRCh38
          GenBankmRNA BI768890.1 - 3 533 Chr. 5 - 112160880 - 112162382 [+] on Build GRCh38
          GenBankmRNA AV688894.1 - 3 252 Chr. 5 - 112161178 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA770717.1 - 3 552 Chr. 5 - 112160865 - 112162235 [+] on Build GRCh38
          GenBankmRNA DA519930.1 - 3 551 Chr. 5 - 112160865 - 112162214 [+] on Build GRCh38
          GenBankmRNA BF969780.1 - 3 251 Chr. 5 - 112161168 - 112162315 [+] on Build GRCh38
          GenBankmRNA BI756062.1 - 3 536 Chr. 5 - 112160880 - 112162306 [+] on Build GRCh38
          GenBankmRNA CF123850.1 - 3 582 Chr. 5 - 112160873 - 112162258 [+] on Build GRCh38
          GenBankmRNA CF127270.1 - 3 510 Chr. 5 - 112160913 - 112162312 [+] on Build GRCh38
          GenBankmRNA AW954951.1 - 3 513 Chr. 5 - 112160902 - 112162298 [+] on Build GRCh38
          GenBankmRNA CN301203.1 - 3 594 Chr. 5 - 112160881 - 112162303 [+] on Build GRCh38
          GenBankmRNA DA782287.1 - 3 553 Chr. 5 - 112160864 - 112162225 [+] on Build GRCh38
          GenBankmRNA DA681460.1 - 3 561 Chr. 5 - 112160866 - 112162231 [+] on Build GRCh38
          GenBankmRNA DB532978.1 - 3 337 Chr. 5 - 112161081 - 112162306 [+] on Build GRCh38
          GenBankmRNA BF967293.2 - 3 557 Chr. 5 - 112160862 - 112162294 [+] on Build GRCh38
          GenBankmRNA BF727429.1 - 3 535 Chr. 5 - 112160883 - 112162307 [+] on Build GRCh38
          GenBankmRNA BF314746.1 - 529 Chr. - - [-] on Build
          GenBankmRNA BM695362.1 - 3 531 Chr. 5 - 112160886 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA076661.1 - 3 552 Chr. 5 - 112160864 - 112162221 [+] on Build GRCh38
          GenBankmRNA DA532218.1 - 3 553 Chr. 5 - 112160864 - 112162250 [+] on Build GRCh38
          GenBankmRNA DA528195.1 - 3 552 Chr. 5 - 112160865 - 112162250 [+] on Build GRCh38
          GenBankmRNA DA121255.1 - 3 544 Chr. 5 - 112160873 - 112162210 [+] on Build GRCh38
          GenBankmRNA BE392562.1 - 3 508 Chr. 5 - 112160905 - 112162305 [+] on Build GRCh38
          GenBankmRNA AL542607.3 - 2 913 Chr. 5 - 112160880 - 112162294 [+] on Build GRCh38
          GenBankmRNA DB269453.1 - 3 553 Chr. 5 - 112160864 - 112162209 [+] on Build GRCh38
          GenBankmRNA DA856584.1 - 3 552 Chr. 5 - 112160865 - 112162209 [+] on Build GRCh38
          GenBankmRNA DA207067.1 - 3 555 Chr. 5 - 112160866 - 112162219 [+] on Build GRCh38
          GenBankmRNA BM781998.1 - 3 552 Chr. 5 - 112160866 - 112162268 [+] on Build GRCh38
          GenBankmRNA DA925618.1 - 3 554 Chr. 5 - 112160865 - 112162232 [+] on Build GRCh38
          GenBankmRNA DA500844.1 - 3 550 Chr. 5 - 112160867 - 112162209 [+] on Build GRCh38
          GenBankmRNA BF791660.1 - 531 Chr. - - [-] on Build
          GenBankmRNA DA502642.1 - 3 554 Chr. 5 - 112160866 - 112162210 [+] on Build GRCh38
          GenBankmRNA BE259773.1 - 3 506 Chr. 5 - 112160911 - 112162310 [+] on Build GRCh38
          GenBankmRNA DA779776.1 - 3 554 Chr. 5 - 112160866 - 112162222 [+] on Build GRCh38
          GenBankmRNA DA210552.1 - 3 551 Chr. 5 - 112160866 - 112162212 [+] on Build GRCh38
          GenBankmRNA BU196182.1 - 536 Chr. - - [-] on Build
          GenBankmRNA DA745136.1 - 3 523 Chr. 5 - 112160896 - 112162227 [+] on Build GRCh38
          GenBankmRNA CD176026.1 - 3 568 Chr. 5 - 112160855 - 112162308 [+] on Build GRCh38
          GenBankmRNA DA287168.1 - 3 524 Chr. 5 - 112160894 - 112162249 [+] on Build GRCh38
          GenBankmRNA BE616318.1 - 504 Chr. - - [-] on Build
          GenBankmRNA AL551513.3 - 3 566 Chr. 5 - 112160851 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA522279.1 - 3 555 Chr. 5 - 112160866 - 112162209 [+] on Build GRCh38
          GenBankmRNA DB171189.1 - 3 552 Chr. 5 - 112160865 - 112162231 [+] on Build GRCh38
          GenBankmRNA BE395909.1 - 3 510 Chr. 5 - 112160905 - 112162306 [+] on Build GRCh38
          GenBankmRNA CD513102.1 - 3 536 Chr. 5 - 112160881 - 112162307 [+] on Build GRCh38
          GenBankmRNA BX115634.1 - 3 390 Chr. 5 - 112161027 - 112162315 [+] on Build GRCh38
          GenBankmRNA AA248772.1 - 3 226 Chr. 5 - 112161192 - 112162298 [+] on Build GRCh38
          GenBankmRNA DA305841.1 - 3 552 Chr. 5 - 112160865 - 112162212 [+] on Build GRCh38
          GenBankmRNA BF314470.1 - 529 Chr. - - [-] on Build
          GenBankmRNA AA648113.1 - 3 387 Chr. 5 - 112161035 - 112162287 [+] on Build GRCh38
          GenBankmRNA CN301200.1 - 3 532 Chr. 5 - 112160885 - 112162303 [+] on Build GRCh38
          GenBankmRNA CB125170.1 - 3 544 Chr. 5 - 112160873 - 112162260 [+] on Build GRCh38
          GenBankmRNA CN301202.1 - 3 516 Chr. 5 - 112160901 - 112162307 [+] on Build GRCh38
          GenBankmRNA AA496650.1 - 3 403 Chr. 5 - 112161027 - 112162277 [+] on Build GRCh38
          GenBankmRNA BP343090.1 - 3 553 Chr. 5 - 112160864 - 112162227 [+] on Build GRCh38
          GenBankmRNA F36336.1 - 282 Chr. - - [-] on Build
          GenBankmRNA DA495234.1 - 3 555 Chr. 5 - 112160866 - 112162226 [+] on Build GRCh38
          GenBankmRNA BM786647.1 - 3 542 Chr. 5 - 112160874 - 112162280 [+] on Build GRCh38
          GenBankmRNA DA393479.1 - 3 552 Chr. 5 - 112160865 - 112162239 [+] on Build GRCh38
          GenBankmRNA BP358751.1 - 3 553 Chr. 5 - 112160864 - 112162220 [+] on Build GRCh38
          GenBankmRNA CN301208.1 - 3 529 Chr. 5 - 112160888 - 112162307 [+] on Build GRCh38
          GenBankmRNA DA902367.1 - 3 552 Chr. 5 - 112160865 - 112162229 [+] on Build GRCh38
          GenBankmRNA DA116174.1 - 3 559 Chr. 5 - 112160860 - 112162226 [+] on Build GRCh38
          GenBankmRNA DA790029.1 - 3 553 Chr. 5 - 112160864 - 112162220 [+] on Build GRCh38
          GenBankmRNA CD248174.1 - 568 Chr. - - [-] on Build
          GenBankmRNA DA260934.1 - 3 552 Chr. 5 - 112160865 - 112162215 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0