Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR133B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029903.1 | 1 | 37 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AA234151.1 | - | 1 | 87 | Chr. 6 - 52148872 - 52149316 [+] on Build GRCh38 |