Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LOC148413 SIRNA Assays ›
          Gene Symbol:
          LOC148413
          Gene Name:
          uncharacterized LOC148413
          Gene Aliases:
          -
          Chromosome Location:
          Chr.1: 1399530 - 1402046 on Build GRCh38
          UniGene ID:
          Hs.741357
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n259655

          Purification
          Size
          Availability Inventoried
          Catalog # 4392420
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LOC148413
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015434.1 2 194
          GenBank mRNA AK024040.1 - 2 168
          AK092572.1 - 1 386
          AK297832.1 - 2 183
          BC068600.1 - 2 151
          BC080546.1 - 2 150
          BC104242.1 - 2 198
          BC104243.1 - 2 194
          BC112998.1 - 2 192
          BC112999.1 - 2 188
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA HY079806.1 - 2 157 Chr. 1 - 1399567 - 1400889 [+] on Build GRCh38
          GenBankmRNA BI118146.1 - 2 136 Chr. 1 - 1399584 - 1401182 [+] on Build GRCh38
          GenBankmRNA AW960090.1 - 2 151 Chr. 1 - 1399573 - 1400906 [+] on Build GRCh38
          GenBankmRNA CX758374.1 - 2 167 Chr. 1 - 1399557 - 1401019 [+] on Build GRCh38
          GenBankmRNA BX376671.2 - 2 175 Chr. 1 - 1399575 - 1401329 [+] on Build GRCh38
          GenBankmRNA DA915106.1 - 2 142 Chr. 1 - 1399582 - 1400901 [+] on Build GRCh38
          GenBankmRNA DA698774.1 - 2 156 Chr. 1 - 1399564 - 1400871 [+] on Build GRCh38
          GenBankmRNA BM544771.1 - 1 586 Chr. 1 - 1399628 - 1400906 [+] on Build GRCh38
          GenBankmRNA BG254416.1 - 2 165 Chr. 1 - 1399555 - 1401146 [+] on Build GRCh38
          GenBankmRNA BE795124.1 - 126 Chr. - - [-] on Build
          GenBankmRNA BQ064163.1 - 130 Chr. - - [-] on Build
          GenBankmRNA AU143273.1 - 2 168 Chr. 1 - 1399552 - 1401048 [+] on Build GRCh38
          GenBankmRNA DA954145.1 - 2 366 Chr. 1 - 1399555 - 1400709 [+] on Build GRCh38
          GenBankmRNA DA931499.1 - 2 137 Chr. 1 - 1399587 - 1400871 [+] on Build GRCh38
          GenBankmRNA AL527639.3 - 2 151 Chr. 1 - 1399575 - 1401270 [+] on Build GRCh38
          GenBankmRNA BM921335.1 - 165 Chr. - - [-] on Build
          GenBankmRNA BX387394.2 - 2 148 Chr. 1 - 1399572 - 1401353 [+] on Build GRCh38
          GenBankmRNA DA254079.1 - 2 168 Chr. 1 - 1399552 - 1400775 [+] on Build GRCh38
          GenBankmRNA BM920213.1 - 1 626 Chr. 1 - 1399583 - 1400832 [+] on Build GRCh38
          GenBankmRNA DA489530.1 - 2 173 Chr. 1 - 1399548 - 1400868 [+] on Build GRCh38
          GenBankmRNA DC393491.1 - 2 174 Chr. 1 - 1399546 - 1400880 [+] on Build GRCh38
          GenBankmRNA BE901076.1 - 2 151 Chr. 1 - 1399573 - 1401019 [+] on Build GRCh38
          GenBankmRNA BE788817.1 - 2 150 Chr. 1 - 1399574 - 1400905 [+] on Build GRCh38
          GenBankmRNA BQ061816.1 - 2 131 Chr. 1 - 1399594 - 1401252 [+] on Build GRCh38
          GenBankmRNA DA988281.1 - 2 168 Chr. 1 - 1399552 - 1400894 [+] on Build GRCh38
          GenBankmRNA DA075321.1 - 2 173 Chr. 1 - 1399552 - 1400781 [+] on Build GRCh38
          GenBankmRNA AL558971.3 - 2 151 Chr. 1 - 1399575 - 1401188 [+] on Build GRCh38
          GenBankmRNA DA209785.1 - 2 172 Chr. 1 - 1399548 - 1400880 [+] on Build GRCh38
          GenBankmRNA BI552758.1 - 2 156 Chr. 1 - 1399564 - 1401134 [+] on Build GRCh38
          GenBankmRNA BQ069125.1 - 175 Chr. - - [-] on Build
          GenBankmRNA H06932.1 - 2 141 Chr. 1 - 1399579 - 1400765 [+] on Build GRCh38
          GenBankmRNA AA430332.1 - 2 124 Chr. 1 - 1399592 - 1400720 [+] on Build GRCh38
          GenBankmRNA DC393094.1 - 2 196 Chr. 1 - 1399529 - 1400835 [+] on Build GRCh38
          GenBankmRNA DA227652.1 - 1 345 Chr. 1 - 1399848 - 1400722 [+] on Build GRCh38
          GenBankmRNA DA274691.1 - 1 342 Chr. 1 - 1399850 - 1400722 [+] on Build GRCh38
          GenBankmRNA HY072670.1 - 2 159 Chr. 1 - 1399564 - 1400811 [+] on Build GRCh38
          GenBankmRNA DB115625.1 - 2 159 Chr. 1 - 1399565 - 1400880 [+] on Build GRCh38
          GenBankmRNA BI767355.1 - 2 138 Chr. 1 - 1399582 - 1401172 [+] on Build GRCh38
          GenBankmRNA AL528291.3 - 2 327 Chr. 1 - 1399572 - 1401180 [+] on Build GRCh38
          GenBankmRNA BM921447.1 - 2 156 Chr. 1 - 1399572 - 1401278 [+] on Build GRCh38
          GenBankmRNA BI911209.1 - 2 233 Chr. 1 - 1399526 - 1401087 [+] on Build GRCh38
          GenBankmRNA BI758740.1 - 2 171 Chr. 1 - 1399550 - 1401296 [+] on Build GRCh38
          GenBankmRNA CR986326.1 - 2 143 Chr. 1 - 1399581 - 1400968 [+] on Build GRCh38
          GenBankmRNA BG328690.1 - 2 152 Chr. 1 - 1399573 - 1401103 [+] on Build GRCh38
          GenBankmRNA DA035496.1 - 2 164 Chr. 1 - 1399556 - 1400802 [+] on Build GRCh38
          GenBankmRNA BQ058973.1 - 2 130 Chr. 1 - 1399594 - 1401350 [+] on Build GRCh38
          GenBankmRNA BI517383.1 - 2 139 Chr. 1 - 1399581 - 1401219 [+] on Build GRCh38
          GenBankmRNA DC339388.1 - 2 179 Chr. 1 - 1399541 - 1400757 [+] on Build GRCh38
          GenBankmRNA DB262233.1 - 2 169 Chr. 1 - 1399552 - 1400902 [+] on Build GRCh38
          GenBankmRNA BM547793.1 - 2 133 Chr. 1 - 1399591 - 1401261 [+] on Build GRCh38
          GenBankmRNA HY074502.1 - 2 157 Chr. 1 - 1399567 - 1400900 [+] on Build GRCh38
          GenBankmRNA DA254561.1 - 2 168 Chr. 1 - 1399552 - 1400857 [+] on Build GRCh38
          GenBankmRNA DT220464.1 - 2 170 Chr. 1 - 1399550 - 1401000 [+] on Build GRCh38
          GenBankmRNA AW957724.1 - 2 91 Chr. 1 - 1399639 - 1400822 [+] on Build GRCh38
          GenBankmRNA AA206537.1 - 2 142 Chr. 1 - 1399578 - 1400851 [+] on Build GRCh38
          GenBankmRNA BF037666.1 - 2 108 Chr. 1 - 1399612 - 1401215 [+] on Build GRCh38
          GenBankmRNA AL543994.3 - 1 619 Chr. 1 - 1399572 - 1400791 [+] on Build GRCh38
          GenBankmRNA DC356520.1 - 2 183 Chr. 1 - 1399541 - 1400879 [+] on Build GRCh38
          GenBankmRNA CD580461.2 - 2 149 Chr. 1 - 1399581 - 1401143 [+] on Build GRCh38
          GenBankmRNA DA141713.1 - 2 157 Chr. 1 - 1399564 - 1400877 [+] on Build GRCh38
          GenBankmRNA DA132942.1 - 2 168 Chr. 1 - 1399552 - 1400872 [+] on Build GRCh38
          GenBankmRNA AL548695.3 - 2 118 Chr. 1 - 1399606 - 1401277 [+] on Build GRCh38
          GenBankmRNA BI835920.1 - 2 152 Chr. 1 - 1399572 - 1401087 [+] on Build GRCh38
          GenBankmRNA BQ070201.1 - 2 148 Chr. 1 - 1399576 - 1401291 [+] on Build GRCh38
          GenBankmRNA BI522617.1 - 2 193 Chr. 1 - 1399527 - 1400972 [+] on Build GRCh38
          GenBankmRNA DA074295.1 - 1 386 Chr. 1 - 1399806 - 1400684 [+] on Build GRCh38
          GenBankmRNA DC381095.1 - 2 179 Chr. 1 - 1399541 - 1400857 [+] on Build GRCh38
          GenBankmRNA BI257793.1 - 65 Chr. - - [-] on Build
          GenBankmRNA DT218114.1 - 2 170 Chr. 1 - 1399550 - 1400923 [+] on Build GRCh38
          GenBankmRNA AL533370.3 - 2 185 Chr. 1 - 1399561 - 1401295 [+] on Build GRCh38
          GenBankmRNA DB246546.1 - 2 175 Chr. 1 - 1399549 - 1400871 [+] on Build GRCh38
          GenBankmRNA DA061626.1 - 2 168 Chr. 1 - 1399552 - 1400898 [+] on Build GRCh38
          GenBankmRNA CX754012.1 - 2 165 Chr. 1 - 1399555 - 1401063 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-59df64f8c5-6gx82:80/100.66.130.62:80.
          git-commit: fe83b6adba5b7aa08a343fc2038c841d76c6a25b
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0