Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other UBA6-AS1 SIRNA Assays ›
          Gene Symbol:
          UBA6-AS1
          Gene Name:
          UBA6 antisense RNA 1 (head to head)
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 67701278 - 67722505 on Build GRCh38
          UniGene ID:
          Hs.479879
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n260013

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: UBA6-AS1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015439.1 2 616
          GenBank mRNA AK093682.1 - 2 533
          AK129519.1 - 2 561
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM919725.1 - 2 549 Chr. 4 - 67701364 - 67721630 [+] on Build GRCh38
          GenBankmRNA BI758691.1 - 562 Chr. - - [-] on Build
          GenBankmRNA DA367673.1 - 2 533 Chr. 4 - 67701361 - 67720864 [+] on Build GRCh38
          GenBankmRNA DA784614.1 - 2 537 Chr. 4 - 67701361 - 67720862 [+] on Build GRCh38
          GenBankmRNA DB256801.1 - 2 533 Chr. 4 - 67701361 - 67720855 [+] on Build GRCh38
          GenBankmRNA CD512487.1 - 2 538 Chr. 4 - 67701358 - 67721024 [+] on Build GRCh38
          GenBankmRNA DA915331.1 - 2 510 Chr. 4 - 67701384 - 67720864 [+] on Build GRCh38
          GenBankmRNA DA278378.1 - 2 533 Chr. 4 - 67701361 - 67720853 [+] on Build GRCh38
          GenBankmRNA BU552754.1 - 2 541 Chr. 4 - 67701353 - 67721161 [+] on Build GRCh38
          GenBankmRNA DA486250.1 - 2 533 Chr. 4 - 67701361 - 67720854 [+] on Build GRCh38
          GenBankmRNA DA032898.1 - 2 561 Chr. 4 - 67701333 - 67720854 [+] on Build GRCh38
          GenBankmRNA DB017974.1 - 2 528 Chr. 4 - 67701367 - 67720872 [+] on Build GRCh38
          GenBankmRNA DA627363.1 - 2 533 Chr. 4 - 67701361 - 67720841 [+] on Build GRCh38
          GenBankmRNA DA509740.1 - 2 535 Chr. 4 - 67701359 - 67720860 [+] on Build GRCh38
          GenBankmRNA DA381717.1 - 2 519 Chr. 4 - 67701377 - 67720879 [+] on Build GRCh38
          GenBankmRNA BM808997.1 - 617 Chr. - - [-] on Build
          GenBankmRNA DB182975.1 - 2 533 Chr. 4 - 67701361 - 67720871 [+] on Build GRCh38
          GenBankmRNA DA499519.1 - 2 561 Chr. 4 - 67701333 - 67720845 [+] on Build GRCh38
          GenBankmRNA DA151514.1 - 2 536 Chr. 4 - 67701361 - 67720848 [+] on Build GRCh38
          GenBankmRNA DB197211.1 - 2 533 Chr. 4 - 67701361 - 67720873 [+] on Build GRCh38
          GenBankmRNA DB106399.1 - 2 533 Chr. 4 - 67701361 - 67720863 [+] on Build GRCh38
          GenBankmRNA BM464109.1 - 2 523 Chr. 4 - 67701383 - 67721702 [+] on Build GRCh38
          GenBankmRNA BI756743.1 - 2 549 Chr. 4 - 67701345 - 67721618 [+] on Build GRCh38
          GenBankmRNA DA509107.1 - 2 533 Chr. 4 - 67701361 - 67720872 [+] on Build GRCh38
          GenBankmRNA DA419829.1 - 2 533 Chr. 4 - 67701361 - 67720857 [+] on Build GRCh38
          GenBankmRNA CA336548.1 - 2 565 Chr. 4 - 67701330 - 67720856 [+] on Build GRCh38
          GenBankmRNA BQ549970.1 - 2 519 Chr. 4 - 67701382 - 67720917 [+] on Build GRCh38
          GenBankmRNA DA619761.1 - 2 561 Chr. 4 - 67701333 - 67720863 [+] on Build GRCh38
          GenBankmRNA BU528383.1 - 2 516 Chr. 4 - 67701378 - 67721161 [+] on Build GRCh38
          GenBankmRNA DB095559.1 - 2 533 Chr. 4 - 67701361 - 67720851 [+] on Build GRCh38
          GenBankmRNA DB028535.1 - 2 518 Chr. 4 - 67701377 - 67720905 [+] on Build GRCh38
          GenBankmRNA DA527886.1 - 2 561 Chr. 4 - 67701333 - 67720844 [+] on Build GRCh38
          GenBankmRNA DA481857.1 - 2 476 Chr. 4 - 67701419 - 67720928 [+] on Build GRCh38
          GenBankmRNA DA424108.1 - 2 483 Chr. 4 - 67701411 - 67720906 [+] on Build GRCh38
          GenBankmRNA CK820330.1 - 2 519 Chr. 4 - 67701382 - 67720886 [+] on Build GRCh38
          GenBankmRNA BX383582.2 - 2 539 Chr. 4 - 67701355 - 67721063 [+] on Build GRCh38
          GenBankmRNA DA965517.1 - 2 533 Chr. 4 - 67701361 - 67720863 [+] on Build GRCh38
          GenBankmRNA BM919337.1 - 2 502 Chr. 4 - 67701412 - 67721274 [+] on Build GRCh38
          GenBankmRNA DB105850.1 - 2 533 Chr. 4 - 67701361 - 67720852 [+] on Build GRCh38
          GenBankmRNA DA697725.1 - 2 512 Chr. 4 - 67701382 - 67720891 [+] on Build GRCh38
          GenBankmRNA BM021370.1 - 2 534 Chr. 4 - 67701360 - 67720854 [+] on Build GRCh38
          GenBankmRNA DA876853.1 - 2 533 Chr. 4 - 67701361 - 67720882 [+] on Build GRCh38
          GenBankmRNA DA145708.1 - 2 536 Chr. 4 - 67701358 - 67720856 [+] on Build GRCh38
          GenBankmRNA BE886090.1 - 2 493 Chr. 4 - 67701401 - 67721071 [+] on Build GRCh38
          GenBankmRNA CN369887.1 - 2 484 Chr. 4 - 67701410 - 67720913 [+] on Build GRCh38
          GenBankmRNA DB106574.1 - 2 533 Chr. 4 - 67701361 - 67720860 [+] on Build GRCh38
          GenBankmRNA DA068004.1 - 2 562 Chr. 4 - 67701330 - 67720844 [+] on Build GRCh38
          GenBankmRNA CD108502.1 - 2 507 Chr. 4 - 67701414 - 67721210 [+] on Build GRCh38
          GenBankmRNA DB169923.1 - 2 560 Chr. 4 - 67701334 - 67720847 [+] on Build GRCh38
          GenBankmRNA DA251354.1 - 2 533 Chr. 4 - 67701361 - 67720875 [+] on Build GRCh38
          GenBankmRNA DA930876.1 - 2 483 Chr. 4 - 67701411 - 67720914 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix