Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other IP6K2 SIRNA Assays ›
          Gene Symbol:
          IP6K2
          Gene Name:
          inositol hexakisphosphate kinase 2
          Gene Aliases:
          IHPK2, PIUS
          Chromosome Location:
          Chr.3: 48688003 - 48717278 on Build GRCh38
          UniGene ID:
          Hs.595983
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n261503

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: IP6K2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_027437.2 2, 3 455
          NR_027438.2 2, 3 452
          XM_011533817.1 XP_011532119.1 2, 3 392
          XM_017006587.1 XP_016862076.1 3, 4 582
          GenBank mRNA BC001864.1 - 2, 3 377
          CD248129.1 - 2, 3 418
          CF139656.1 - 2, 3 386
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA DA753143.1 - 2, 3 392 Chr. 3 - 48694695 - 48717214 [-] on Build GRCh38
          GenBankmRNA EL737198.1 - 1, 2 203 Chr. 3 - 48694757 - 48695291 [-] on Build GRCh38
          GenBankmRNA BI549902.1 - 3, 4 568 Chr. 3 - 48694175 - 48717255 [-] on Build GRCh38
          GenBankmRNA BQ653292.1 - 3, 4 495 Chr. 3 - 48694127 - 48717199 [-] on Build GRCh38
          GenBankmRNA BE267366.1 - 2, 3 358 Chr. 3 - 48694071 - 48717184 [-] on Build GRCh38
          GenBankmRNA BQ073372.1 - 2, 3 364 Chr. 3 - 48693862 - 48717189 [-] on Build GRCh38
          GenBankmRNA BI548019.1 - 432 Chr. - - [-] on Build
          GenBankmRNA DA900565.1 - 2, 3 396 Chr. 3 - 48694196 - 48717219 [-] on Build GRCh38
          GenBankmRNA BF308275.1 - 376 Chr. - - [-] on Build
          GenBankmRNA CN362790.1 - 2, 3 349 Chr. 3 - 48694192 - 48717175 [-] on Build GRCh38
          GenBankmRNA BG740735.1 - 2, 3 354 Chr. 3 - 48693830 - 48717180 [-] on Build GRCh38
          GenBankmRNA BM852971.1 - 2, 3 369 Chr. 3 - 48694693 - 48717195 [-] on Build GRCh38
          GenBankmRNA CN362776.1 - 2, 3 382 Chr. 3 - 48694035 - 48717205 [-] on Build GRCh38

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          phosphate ion transport
          negative regulation of cell growth
          positive regulation of apoptotic process
          inositol phosphate metabolic process
          phosphatidylinositol phosphorylation
          type I interferon signaling pathway
          inositol-1,3,4,5,6-pentakisphosphate kinase activity
          inositol hexakisphosphate kinase activity
          inositol heptakisphosphate kinase activity
          inositol hexakisphosphate 5-kinase activity
          protein binding
          ATP binding
          inositol-1,4,5-trisphosphate 3-kinase activity
          inositol hexakisphosphate 1-kinase activity
          inositol hexakisphosphate 3-kinase activity
          inositol 5-diphosphate pentakisphosphate 5-kinase activity
          inositol diphosphate tetrakisphosphate kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-97865cc7b-9fgnx:80/100.66.128.131:80.
          git-commit: d53bc399386730d6c5c6d01fe94b21e0ad3fdbfa
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.33.0-2025.07.30-1.0