Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01106 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027244.1 | 2 | 716 | |||
GenBank mRNA | AK095678.1 | - | 2 | 1025 | ||
CR749351.1 | - | 3 | 807 |
Gene Symbol: LINC01123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046110.1 | 4 | 2132 | |||
NR_046111.1 | 3 | 1947 | ||||
NR_046112.1 | 2 | 705 | ||||
GenBank mRNA | AK056084.1 | - | 4 | 2049 |