Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SNHG17 SIRNA Assays ›
          Gene Symbol:
          SNHG17
          Gene Name:
          small nucleolar RNA host gene 17
          Gene Aliases:
          -
          Chromosome Location:
          Chr.20: 38420592 - 38435375 on Build GRCh38
          UniGene ID:
          Hs.400876
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n263789

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: SNHG17
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_015366.4 7 702
          NR_027241.2 6 815
          GenBank mRNA AK022745.1 - 3 1765
          BC032119.2 - 6 781
          BC052370.1 - 6 416
          BX640887.1 - 5 4821
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BM806173.1 - 2 85 Chr. 20 - 38420592 - 38426471 [-] on Build GRCh38
          GenBankmRNA BM853469.1 - 3 153 Chr. 20 - 38420598 - 38431077 [-] on Build GRCh38
          GenBankmRNA HY111703.1 - 5 444 Chr. 20 - 38421013 - 38435328 [-] on Build GRCh38
          GenBankmRNA BF206691.1 - 5 238 Chr. 20 - 38420596 - 38434566 [-] on Build GRCh38
          GenBankmRNA BQ673916.1 - 5 601 Chr. 20 - 38420679 - 38435316 [-] on Build GRCh38
          GenBankmRNA BF340988.1 - 6 412 Chr. 20 - 38420709 - 38435274 [-] on Build GRCh38
          GenBankmRNA N35265.1 - 3 133 Chr. 20 - 38420807 - 38431057 [-] on Build GRCh38
          GenBankmRNA AA429963.1 - 4 223 Chr. 20 - 38420800 - 38431071 [-] on Build GRCh38
          GenBankmRNA BM562678.1 - 647 Chr. - - [-] on Build
          GenBankmRNA AW629655.1 - 5 403 Chr. 20 - 38421022 - 38434587 [-] on Build GRCh38
          GenBankmRNA BG482037.1 - 5 617 Chr. 20 - 38420710 - 38435331 [-] on Build GRCh38
          GenBankmRNA CV802901.1 - 2 125 Chr. 20 - 38420592 - 38426505 [-] on Build GRCh38
          GenBankmRNA BG106290.1 - 515 Chr. - - [-] on Build
          GenBankmRNA BF311155.1 - 6 434 Chr. 20 - 38420690 - 38435331 [-] on Build GRCh38
          GenBankmRNA BG481678.1 - 6 509 Chr. 20 - 38420597 - 38435277 [-] on Build GRCh38
          GenBankmRNA BG481329.1 - 6 511 Chr. 20 - 38420625 - 38435277 [-] on Build GRCh38
          GenBankmRNA BG330525.1 - 6 531 Chr. 20 - 38420726 - 38435341 [-] on Build GRCh38
          GenBankmRNA BU145155.1 - 5 421 Chr. 20 - 38420641 - 38435306 [-] on Build GRCh38
          GenBankmRNA CB997412.1 - 486 Chr. - - [-] on Build
          GenBankmRNA BU500384.1 - 7 606 Chr. 20 - 38420792 - 38435297 [-] on Build GRCh38
          GenBankmRNA BE408029.1 - 7 490 Chr. 20 - 38420877 - 38435258 [-] on Build GRCh38
          GenBankmRNA BG754976.1 - 5 214 Chr. 20 - 38420592 - 38434528 [-] on Build GRCh38
          GenBankmRNA BU196029.1 - 5 466 Chr. 20 - 38420644 - 38435351 [-] on Build GRCh38
          GenBankmRNA BG106683.1 - 5 447 Chr. 20 - 38420653 - 38435333 [-] on Build GRCh38
          GenBankmRNA DB541814.1 - 2 96 Chr. 20 - 38420590 - 38426483 [-] on Build GRCh38
          GenBankmRNA BI754333.1 - 5 428 Chr. 20 - 38420592 - 38435314 [-] on Build GRCh38
          GenBankmRNA BI836626.1 - 7 569 Chr. 20 - 38420804 - 38435263 [-] on Build GRCh38
          GenBankmRNA BX363714.2 - 7 686 Chr. 20 - 38420663 - 38435303 [-] on Build GRCh38
          GenBankmRNA BI194992.1 - 6 434 Chr. 20 - 38420746 - 38435331 [-] on Build GRCh38
          GenBankmRNA BQ669163.1 - 5 601 Chr. 20 - 38420721 - 38435316 [-] on Build GRCh38
          GenBankmRNA EL953190.1 - 5 410 Chr. 20 - 38420773 - 38434566 [-] on Build GRCh38
          GenBankmRNA BI084345.1 - 5 603 Chr. 20 - 38420793 - 38435317 [-] on Build GRCh38
          GenBankmRNA BI195102.1 - 6 433 Chr. 20 - 38421024 - 38435331 [-] on Build GRCh38
          GenBankmRNA BU153111.1 - 634 Chr. - - [-] on Build
          GenBankmRNA BG292447.1 - 5 408 Chr. 20 - 38420575 - 38435294 [-] on Build GRCh38
          GenBankmRNA BG336608.1 - 462 Chr. - - [-] on Build
          GenBankmRNA CD675826.1 - 6 412 Chr. 20 - 38420829 - 38435310 [-] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix