Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LINC00301 SIRNA Assays ›
          Gene Symbol:
          LINC00301
          Gene Name:
          long intergenic non-protein coding RNA 301
          Gene Aliases:
          C11orf64, NCRNA00301
          Chromosome Location:
          Chr.11: 60615751 - 60687149 on Build GRCh38
          UniGene ID:
          Hs.372650
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID n264956

          Purification
          Size
          Availability Made To Order
          Catalog # 4390771
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: LINC00301
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NR_026946.1 3 225
          GenBank mRNA AK058123.1 - 3 225
          BC029583.1 - 3 203
          BX103095.1 - 3 206
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
          GenBankmRNA BU852641.1 - 222 Chr. - - [-] on Build
          GenBankmRNA FY211930.1 - 154 Chr. - - [-] on Build
          GenBankmRNA BI463377.1 - 3 208 Chr. 11 - 60615769 - 60687149 [+] on Build GRCh38
          GenBankmRNA BU561608.1 - 3 225 Chr. 11 - 60615752 - 60701037 [+] on Build GRCh38
          GenBankmRNA FY212023.1 - 3 156 Chr. 11 - 60615820 - 60700969 [+] on Build GRCh38
          GenBankmRNA BI458881.1 - 3 235 Chr. 11 - 60615743 - 60687140 [+] on Build GRCh38
          GenBankmRNA HY037540.1 - 3 208 Chr. 11 - 60615769 - 60649285 [+] on Build GRCh38
          GenBankmRNA FY211044.1 - 156 Chr. - - [-] on Build
          GenBankmRNA FY211777.1 - 156 Chr. - - [-] on Build
          GenBankmRNA HY023099.1 - 3 231 Chr. 11 - 60615746 - 60649319 [+] on Build GRCh38
          GenBankmRNA FY210852.1 - 155 Chr. - - [-] on Build
          GenBankmRNA HY027139.1 - 3 231 Chr. 11 - 60615746 - 60649269 [+] on Build GRCh38
          GenBankmRNA FY211272.1 - 156 Chr. - - [-] on Build
          GenBankmRNA DB046498.1 - 3 241 Chr. 11 - 60615736 - 60686939 [+] on Build GRCh38
          GenBankmRNA HY042264.1 - 3 218 Chr. 11 - 60615758 - 60660758 [+] on Build GRCh38
          GenBankmRNA BU854253.1 - 218 Chr. - - [-] on Build
          GenBankmRNA FY213231.1 - 156 Chr. - - [-] on Build
          GenBankmRNA HY013873.1 - 3 226 Chr. 11 - 60615752 - 60653748 [+] on Build GRCh38
          GenBankmRNA HY019916.1 - 3 230 Chr. 11 - 60615746 - 60649298 [+] on Build GRCh38
          GenBankmRNA HY028992.1 - 3 226 Chr. 11 - 60615751 - 60649314 [+] on Build GRCh38

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5bbdb6fccc-rsc2d:80/100.66.130.62:80.
          git-commit: 26de9a04ea7719c3b15362b12d28d672cac851f3
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: feature/2.32.0/CFGMAGR/CFGMAGR-4285/CFGMAGR-4312-Unified_Card_Service_Magellan_Search_Fix