Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD18DP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003291.1 | 10 | 2782 | |||
GenBank mRNA | AK125940.1 | - | 1 | 4835 |
Gene Symbol: ANKRD18A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017014577.1 | XP_016870066.1 | 11 | 3161 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CR596617.1 | - | 1 | 1157 | Chr. 9 - 38594541 - 38595815 [-] on Build GRCh38 |