Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC01128 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_047519.1 | 5 | 709 | |||
NR_047521.1 | 4 | 556 | ||||
NR_047524.1 | 3 | 372 | ||||
NR_047525.1 | 4 | 423 | ||||
GenBank mRNA | AK026873.1 | - | 5 | 601 | ||
AK311732.1 | - | 4 | 459 | |||
BC042880.1 | - | 4 | 539 | |||
BC102012.1 | - | 5 | 584 | |||
BC102023.1 | - | 5 | 584 | |||
BC107481.1 | - | 3 | 247 | |||
BM459718.1 | - | 5 | 582 | |||
CN361275.1 | - | 4 | 562 | |||
DA223705.1 | - | 4 | 423 | |||
DB282323.1 | - | 4 | 459 | |||
DR005973.1 | - | 5 | 621 |