Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM193B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_024019.3 | 7 | 2689 | |||
XM_006714884.1 | XP_006714947.1 | 3 | 279 | |||
XM_011534582.1 | XP_011532884.1 | 6 | 2129 | |||
XM_011534583.1 | XP_011532885.1 | 6 | 1870 | |||
GenBank mRNA | AB067518.1 | - | 7 | 2646 | ||
AK096032.1 | - | 5 | 841 | |||
BC008784.2 | - | 3 | 279 | 3626782 | ||
BC086859.1 | - | 3 | 503 | |||
BX647587.1 | - | 4 | 774 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | DB119531.1 | - | 2 | 201 | Chr. 5 - 177531324 - 177532526 [-] on Build GRCh38 |