Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAPT1-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027696.1 | 1 | 414 | |||
NR_027697.1 | 1 | 414 | ||||
GenBank mRNA | AK093550.1 | - | 1 | 37 | ||
AK096972.1 | - | 1 | 414 | |||
BC132927.1 | - | 1 | 414 | |||
BF732723.1 | - | 1 | 88 | |||
DA640966.1 | - | 1 | 37 | |||
DR004294.1 | - | 1 | 398 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ285984.1 | - | 1 | 48 | Chr. 4 - 16227029 - 16256594 [+] on Build GRCh38 |