Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MIR223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029637.1 | 1 | 73 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV712697.1 | - | 2 | 159 | Chr. X - 66017150 - 66019427 [+] on Build GRCh38 |