Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC284788 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027089.2 | 1 | 585 | |||
NR_027090.1 | 1 | 585 | ||||
GenBank mRNA | BC033532.1 | - | 1 | 585 | ||
BC144090.1 | - | 1 | 457 |