Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027428.1 | 6 | 1396 | |||
NR_027429.1 | 5 | 1309 | ||||
NR_027430.1 | 5 | 1185 | ||||
NR_027431.2 | 5 | 1159 | ||||
NR_027432.1 | 4 | 1072 | ||||
GenBank mRNA | AK021673.1 | - | 5 | 1028 | ||
AK096758.1 | - | 6 | 1098 | |||
AK308811.1 | - | 4 | 625 | |||
AK309654.1 | - | 5 | 808 | |||
AK310261.1 | - | 4 | 762 | |||
BC028046.1 | - | 4 | 733 |