Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM254-AS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027428.1 | 6 | 1705 | |||
NR_027429.1 | 5 | 1618 | ||||
NR_027430.1 | 5 | 1494 | ||||
NR_027431.2 | 5 | 1468 | ||||
NR_027432.1 | 4 | 1381 | ||||
GenBank mRNA | AK021673.1 | - | 5 | 1337 | ||
AK096758.1 | - | 6 | 1407 | |||
AK308811.1 | - | 4 | 934 | |||
BC028046.1 | - | 4 | 1042 |